Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10941671-10941896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEXI
Entrez Gene ID
28955
Gene Name
Dexi homolog
Gene Aliases
MYLE
UniGene
Hs.592051
Gene Chromosome Location
Chr.16: 10928891-10943007 on Build GRCh38
Transcripts
NM_014015.3 XM_011522455.2 XM_017023171.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3944348 c.261C>T p.L87L Chr.16: 10941745-10941745 on Build GRCh38
702004 c.121C>T p.L41L Chr.16: 10941885-10941885 on Build GRCh38
6406814 c.193C>T p.Q65* Chr.16: 10941813-10941813 on Build GRCh38
3817156 c.234G>A p.S78S Chr.16: 10941772-10941772 on Build GRCh38
7232000 c.240T>C p.L80L Chr.16: 10941766-10941766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765635147 Chr.16: 10941822-10941822 on Build GRCh38
rs781328031 Chr.16: 10941840-10941840 on Build GRCh38
rs768372760 Chr.16: 10941806-10941806 on Build GRCh38
rs761510424 Chr.16: 10941809-10941809 on Build GRCh38
rs755249811 Chr.16: 10941781-10941781 on Build GRCh38
rs558374201 Chr.16: 10941708-10941708 on Build GRCh38
rs754123667 Chr.16: 10941697-10941697 on Build GRCh38
rs750751772 Chr.16: 10941835-10941835 on Build GRCh38
rs972512520 Chr.16: 10941804-10941804 on Build GRCh38
rs759542243 Chr.16: 10941734-10941734 on Build GRCh38
rs758438831 Chr.16: 10941838-10941838 on Build GRCh38
rs922461783 Chr.16: 10941842-10941842 on Build GRCh38
rs763479233 Chr.16: 10941836-10941836 on Build GRCh38
rs754254130 Chr.16: 10941756-10941756 on Build GRCh38
rs770699316 Chr.16: 10941789-10941789 on Build GRCh38
rs770914330 Chr.16: 10941725-10941725 on Build GRCh38
rs146324480 Chr.16: 10941780-10941780 on Build GRCh38
rs760417209 Chr.16: 10941751-10941751 on Build GRCh38
rs772106642 Chr.16: 10941799-10941799 on Build GRCh38
rs142045151 Chr.16: 10941716-10941716 on Build GRCh38
rs745850724 Chr.16: 10941798-10941798 on Build GRCh38
rs763616342 Chr.16: 10941754-10941754 on Build GRCh38
rs978451134 Chr.16: 10941752-10941752 on Build GRCh38
rs958548331 Chr.16: 10941777-10941782 on Build GRCh38
rs745621400 Chr.16: 10941867-10941867 on Build GRCh38
rs758536904 Chr.16: 10941711-10941711 on Build GRCh38
rs776085726 Chr.16: 10941808-10941808 on Build GRCh38
rs766820112 Chr.16: 10941837-10941837 on Build GRCh38
rs752920646 Chr.16: 10941846-10941846 on Build GRCh38
rs537643712 Chr.16: 10941730-10941730 on Build GRCh38
rs758877371 Chr.16: 10941779-10941779 on Build GRCh38
rs757816732 Chr.16: 10941698-10941698 on Build GRCh38
rs1022570666 Chr.16: 10941747-10941747 on Build GRCh38
rs778838339 Chr.16: 10941794-10941794 on Build GRCh38
rs760058332 Chr.16: 10941803-10941803 on Build GRCh38
rs896064204 Chr.16: 10941724-10941724 on Build GRCh38
rs751650005 Chr.16: 10941838-10941838 on Build GRCh38
rs771767260 Chr.16: 10941741-10941741 on Build GRCh38
rs756338689 Chr.16: 10941853-10941853 on Build GRCh38
rs770786777 Chr.16: 10941855-10941855 on Build GRCh38
rs750982081 Chr.16: 10941773-10941773 on Build GRCh38
rs920602929 Chr.16: 10941844-10941844 on Build GRCh38
rs534448508 Chr.16: 10941700-10941700 on Build GRCh38
rs781212436 Chr.16: 10941785-10941785 on Build GRCh38
rs778893300 Chr.16: 10941856-10941856 on Build GRCh38
rs775119065 Chr.16: 10941742-10941742 on Build GRCh38
rs571916203 Chr.16: 10941717-10941717 on Build GRCh38
rs768726996 Chr.16: 10941713-10941713 on Build GRCh38
rs750690209 Chr.16: 10941794-10941794 on Build GRCh38
rs976621499 Chr.16: 10941787-10941787 on Build GRCh38
rs747276966 Chr.16: 10941712-10941712 on Build GRCh38
rs762030120 Chr.16: 10941763-10941763 on Build GRCh38
rs765782979 Chr.16: 10941769-10941769 on Build GRCh38
rs374035200 Chr.16: 10941800-10941800 on Build GRCh38

View Full Product Details