Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 172809306-172809577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNTL6
Entrez Gene ID
442117
Gene Name
polypeptide N-acetylgalactosaminyltransferase-like 6
Gene Aliases
GALNACT20, GALNT17, GalNAc-T6L
UniGene
Hs.386236
Gene Chromosome Location
Chr.4: 171812690-173040408 on Build GRCh38
Transcripts
NM_001034845.2 XM_011531993.2 XM_011531994.1 XM_011531995.1 XM_011531996.1 XM_011531997.1 XM_017008243.1 XM_017008244.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5443293 c.590G>A p.R197Q Chr.4: 172809397-172809397 on Build GRCh38
4555133 c.642G>A p.R214R Chr.4: 172809449-172809449 on Build GRCh38
5418312 c.699C>A p.S233S Chr.4: 172809506-172809506 on Build GRCh38
1671437 c.626G>T p.R209I Chr.4: 172809433-172809433 on Build GRCh38
1053233 c.682C>A p.L228M Chr.4: 172809489-172809489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779482222 Chr.4: 172809554-172809554 on Build GRCh38
rs746865494 Chr.4: 172809543-172809543 on Build GRCh38
rs770998629 Chr.4: 172809558-172809558 on Build GRCh38
rs201908027 Chr.4: 172809458-172809458 on Build GRCh38
rs891843933 Chr.4: 172809397-172809397 on Build GRCh38
rs146548110 Chr.4: 172809448-172809448 on Build GRCh38
rs201336562 Chr.4: 172809546-172809546 on Build GRCh38
rs745628743 Chr.4: 172809384-172809384 on Build GRCh38
rs760632165 Chr.4: 172809396-172809396 on Build GRCh38
rs779980183 Chr.4: 172809539-172809539 on Build GRCh38
rs755415517 Chr.4: 172809341-172809341 on Build GRCh38
rs774170031 Chr.4: 172809447-172809447 on Build GRCh38
rs866548368 Chr.4: 172809426-172809428 on Build GRCh38
rs903698184 Chr.4: 172809485-172809485 on Build GRCh38
rs761810857 Chr.4: 172809328-172809329 on Build GRCh38
rs778309701 Chr.4: 172809517-172809517 on Build GRCh38
rs748054362 Chr.4: 172809553-172809553 on Build GRCh38
rs770486969 Chr.4: 172809383-172809383 on Build GRCh38
rs763130885 Chr.4: 172809421-172809421 on Build GRCh38
rs781555700 Chr.4: 172809345-172809345 on Build GRCh38
rs962290602 Chr.4: 172809534-172809534 on Build GRCh38
rs369789872 Chr.4: 172809328-172809328 on Build GRCh38
rs189797609 Chr.4: 172809454-172809454 on Build GRCh38
rs377525077 Chr.4: 172809393-172809393 on Build GRCh38
rs756840694 Chr.4: 172809512-172809512 on Build GRCh38
rs1000243936 Chr.4: 172809498-172809498 on Build GRCh38
rs745457125 Chr.4: 172809521-172809521 on Build GRCh38
rs759704847 Chr.4: 172809449-172809449 on Build GRCh38
rs756603346 Chr.4: 172809464-172809464 on Build GRCh38
rs143539644 Chr.4: 172809520-172809520 on Build GRCh38
rs756622111 Chr.4: 172809355-172809355 on Build GRCh38
rs72998183 Chr.4: 172809337-172809337 on Build GRCh38
rs371201365 Chr.4: 172809413-172809413 on Build GRCh38
rs778319684 Chr.4: 172809471-172809471 on Build GRCh38
rs141284095 Chr.4: 172809533-172809533 on Build GRCh38
rs374762409 Chr.4: 172809443-172809443 on Build GRCh38
rs199765980 Chr.4: 172809329-172809329 on Build GRCh38
rs1008920854 Chr.4: 172809518-172809518 on Build GRCh38
rs1050541867 Chr.4: 172809506-172809506 on Build GRCh38
rs144181630 Chr.4: 172809390-172809390 on Build GRCh38
rs900659227 Chr.4: 172809459-172809459 on Build GRCh38
rs753166872 Chr.4: 172809347-172809347 on Build GRCh38
rs776755864 Chr.4: 172809549-172809549 on Build GRCh38
rs768770395 Chr.4: 172809410-172809410 on Build GRCh38
rs765309073 Chr.4: 172809463-172809472 on Build GRCh38
rs748857500 Chr.4: 172809382-172809382 on Build GRCh38
rs777209311 Chr.4: 172809376-172809376 on Build GRCh38
rs1011629190 Chr.4: 172809420-172809420 on Build GRCh38
rs766485386 Chr.4: 172809426-172809426 on Build GRCh38
rs753258036 Chr.4: 172809477-172809477 on Build GRCh38

View Full Product Details