Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50891289-50891548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADNP
Entrez Gene ID
23394
Gene Name
activity dependent neuroprotector homeobox
Gene Aliases
ADNP1, HVDAS, MRD28
UniGene
Hs.293736
Gene Chromosome Location
Chr.20: 50888918-50934984 on Build GRCh38
Transcripts
NM_001282531.1 NM_001282532.1 NM_015339.3 NM_181442.2 XM_011528747.2 XM_011528748.2 XM_017027757.1 XM_017027758.1 XM_017027759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1027961 c.3221G>T p.G1074V Chr.20: 50891493-50891493 on Build GRCh38
2762673 c.3166C>T p.R1056C Chr.20: 50891548-50891548 on Build GRCh38
6195939 c.3208G>A p.D1070N Chr.20: 50891506-50891506 on Build GRCh38
2762672 c.3185T>C p.I1062T Chr.20: 50891529-50891529 on Build GRCh38
2762670 c.3244G>C p.D1082H Chr.20: 50891470-50891470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766506206 Chr.20: 50891378-50891378 on Build GRCh38
rs749829736 Chr.20: 50891453-50891453 on Build GRCh38
rs761240 Chr.20: 50891355-50891355 on Build GRCh38
rs200797590 Chr.20: 50891486-50891486 on Build GRCh38
rs199615568 Chr.20: 50891431-50891431 on Build GRCh38
rs765065640 Chr.20: 50891399-50891399 on Build GRCh38
rs764039253 Chr.20: 50891467-50891469 on Build GRCh38
rs763395878 Chr.20: 50891362-50891362 on Build GRCh38
rs1021484823 Chr.20: 50891346-50891346 on Build GRCh38
rs773520278 Chr.20: 50891452-50891452 on Build GRCh38
rs34851327 Chr.20: 50891397-50891398 on Build GRCh38
rs767564900 Chr.20: 50891473-50891473 on Build GRCh38
rs869320662 Chr.20: 50891434-50891435 on Build GRCh38
rs182530989 Chr.20: 50891327-50891327 on Build GRCh38
rs767616705 Chr.20: 50891390-50891390 on Build GRCh38
rs762815251 Chr.20: 50891384-50891386 on Build GRCh38
rs187932970 Chr.20: 50891350-50891350 on Build GRCh38
rs750211598 Chr.20: 50891481-50891481 on Build GRCh38
rs770355320 Chr.20: 50891354-50891354 on Build GRCh38
rs142247083 Chr.20: 50891435-50891435 on Build GRCh38
rs768409843 Chr.20: 50891356-50891356 on Build GRCh38
rs779655684 Chr.20: 50891423-50891423 on Build GRCh38
rs753750584 Chr.20: 50891394-50891394 on Build GRCh38
rs192416296 Chr.20: 50891351-50891351 on Build GRCh38
rs201786178 Chr.20: 50891455-50891455 on Build GRCh38
rs780706686 Chr.20: 50891433-50891433 on Build GRCh38
rs184463340 Chr.20: 50891388-50891388 on Build GRCh38
rs765155352 Chr.20: 50891480-50891480 on Build GRCh38
rs751149237 Chr.20: 50891508-50891508 on Build GRCh38
rs758371438 Chr.20: 50891411-50891411 on Build GRCh38
rs974173754 Chr.20: 50891393-50891393 on Build GRCh38
rs766214850 Chr.20: 50891507-50891507 on Build GRCh38
rs754719375 Chr.20: 50891429-50891429 on Build GRCh38
rs746480446 Chr.20: 50891425-50891425 on Build GRCh38
rs868512194 Chr.20: 50891392-50891392 on Build GRCh38
rs1023455516 Chr.20: 50891475-50891475 on Build GRCh38
rs959731070 Chr.20: 50891370-50891370 on Build GRCh38
rs142825371 Chr.20: 50891387-50891387 on Build GRCh38
rs750407672 Chr.20: 50891410-50891410 on Build GRCh38
rs758410078 Chr.20: 50891503-50891503 on Build GRCh38
rs1011384387 Chr.20: 50891308-50891308 on Build GRCh38
rs759526947 Chr.20: 50891468-50891468 on Build GRCh38
rs960404692 Chr.20: 50891514-50891514 on Build GRCh38
rs999160748 Chr.20: 50891375-50891375 on Build GRCh38
rs147399432 Chr.20: 50891478-50891478 on Build GRCh38
rs775867459 Chr.20: 50891477-50891477 on Build GRCh38
rs770161155 Chr.20: 50891440-50891440 on Build GRCh38
rs774543782 Chr.20: 50891456-50891456 on Build GRCh38

View Full Product Details