Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95625309-95625534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH18A1
Entrez Gene ID
5832
Gene Name
aldehyde dehydrogenase 18 family member A1
Gene Aliases
ADCL3, ARCL3A, GSAS, P5CS, PYCS, SPG9A, SPG9B
UniGene
Hs.500645
Gene Chromosome Location
Chr.10: 95605929-95656810 on Build GRCh38
Transcripts
NM_001017423.1 NM_001323412.1 NM_001323413.1 NM_001323414.1 NM_001323415.1 NM_001323416.1 NM_001323417.1 NM_001323418.1 NM_001323419.1 NM_002860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6598163 c.1199G>A p.R400H Chr.10: 95625409-95625409 on Build GRCh38
6598164 c.1246+1G>A p.? Chr.10: 95625361-95625361 on Build GRCh38
6348175 c.1157C>A p.A386E Chr.10: 95625451-95625451 on Build GRCh38
6275655 c.1230C>A p.D410E Chr.10: 95625378-95625378 on Build GRCh38
921816 c.1183C>A p.L395M Chr.10: 95625425-95625425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781279701 Chr.10: 95625401-95625401 on Build GRCh38
rs376382998 Chr.10: 95625347-95625347 on Build GRCh38
rs758828421 Chr.10: 95625375-95625375 on Build GRCh38
rs751889438 Chr.10: 95625490-95625490 on Build GRCh38
rs940452614 Chr.10: 95625412-95625412 on Build GRCh38
rs767127350 Chr.10: 95625466-95625466 on Build GRCh38
rs769823561 Chr.10: 95625418-95625418 on Build GRCh38
rs775016180 Chr.10: 95625467-95625467 on Build GRCh38
rs767097515 Chr.10: 95625339-95625339 on Build GRCh38
rs770688743 Chr.10: 95625431-95625431 on Build GRCh38
rs370680325 Chr.10: 95625468-95625468 on Build GRCh38
rs777958250 Chr.10: 95625476-95625477 on Build GRCh38
rs755969933 Chr.10: 95625429-95625429 on Build GRCh38
rs369996181 Chr.10: 95625335-95625335 on Build GRCh38
rs767028746 Chr.10: 95625489-95625489 on Build GRCh38
rs747402980 Chr.10: 95625386-95625386 on Build GRCh38
rs554598542 Chr.10: 95625480-95625480 on Build GRCh38
rs759279790 Chr.10: 95625437-95625437 on Build GRCh38
rs755190719 Chr.10: 95625391-95625391 on Build GRCh38
rs748202426 Chr.10: 95625417-95625417 on Build GRCh38
rs774081826 Chr.10: 95625436-95625436 on Build GRCh38
rs890059153 Chr.10: 95625488-95625488 on Build GRCh38
rs1033294741 Chr.10: 95625508-95625508 on Build GRCh38
rs755580308 Chr.10: 95625354-95625354 on Build GRCh38
rs1000034476 Chr.10: 95625507-95625507 on Build GRCh38
rs780616492 Chr.10: 95625383-95625383 on Build GRCh38
rs10509688 Chr.10: 95625485-95625485 on Build GRCh38
rs147348068 Chr.10: 95625371-95625371 on Build GRCh38
rs373405092 Chr.10: 95625336-95625336 on Build GRCh38
rs773131152 Chr.10: 95625427-95625427 on Build GRCh38
rs907590719 Chr.10: 95625410-95625410 on Build GRCh38
rs765778366 Chr.10: 95625361-95625361 on Build GRCh38
rs773959742 Chr.10: 95625331-95625331 on Build GRCh38
rs138114468 Chr.10: 95625472-95625472 on Build GRCh38

View Full Product Details