Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82244662-82244909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1D
Entrez Gene ID
1453
Gene Name
casein kinase 1 delta
Gene Aliases
ASPS, CKIdelta, FASPS2, HCKID
UniGene
Hs.631725
Gene Chromosome Location
Chr.17: 82239317-82273742 on Build GRCh38
Transcripts
NM_001893.4 NM_139062.2 NR_110578.1 XM_005256337.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3989443 c.1230G>A p.Q410Q Chr.17: 82244799-82244799 on Build GRCh38
5076795 c.1213G>A p.A405T Chr.17: 82244816-82244816 on Build GRCh38
5055896 c.1236C>T p.V412V Chr.17: 82244793-82244793 on Build GRCh38
1750423 c.1232C>T p.S411F Chr.17: 82244797-82244797 on Build GRCh38
7258119 c.1218C>T p.S406S Chr.17: 82244811-82244811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs56124628 Chr.17: 82244828-82244828 on Build GRCh38
rs531614835 Chr.17: 82244686-82244686 on Build GRCh38
rs987909717 Chr.17: 82244853-82244853 on Build GRCh38
rs372063084 Chr.17: 82244792-82244792 on Build GRCh38
rs955646124 Chr.17: 82244772-82244772 on Build GRCh38
rs766713067 Chr.17: 82244853-82244854 on Build GRCh38
rs1052350074 Chr.17: 82244852-82244852 on Build GRCh38
rs780889800 Chr.17: 82244755-82244755 on Build GRCh38
rs771974980 Chr.17: 82244786-82244786 on Build GRCh38
rs760928529 Chr.17: 82244793-82244793 on Build GRCh38
rs923406671 Chr.17: 82244776-82244776 on Build GRCh38
rs1004301719 Chr.17: 82244725-82244725 on Build GRCh38
rs375145901 Chr.17: 82244825-82244825 on Build GRCh38
rs369605736 Chr.17: 82244739-82244739 on Build GRCh38
rs754293657 Chr.17: 82244815-82244815 on Build GRCh38
rs200232591 Chr.17: 82244745-82244745 on Build GRCh38
rs956365830 Chr.17: 82244843-82244843 on Build GRCh38
rs375684280 Chr.17: 82244816-82244816 on Build GRCh38
rs1028554074 Chr.17: 82244767-82244767 on Build GRCh38
rs762015722 Chr.17: 82244785-82244785 on Build GRCh38
rs986726708 Chr.17: 82244821-82244821 on Build GRCh38
rs551846940 Chr.17: 82244780-82244780 on Build GRCh38
rs755726288 Chr.17: 82244855-82244855 on Build GRCh38
rs762193663 Chr.17: 82244773-82244774 on Build GRCh38
rs750836786 Chr.17: 82244850-82244852 on Build GRCh38
rs756795818 Chr.17: 82244836-82244836 on Build GRCh38
rs202062937 Chr.17: 82244880-82244880 on Build GRCh38
rs769591591 Chr.17: 82244768-82244768 on Build GRCh38
rs749256944 Chr.17: 82244777-82244777 on Build GRCh38
rs778909401 Chr.17: 82244878-82244878 on Build GRCh38
rs774136610 Chr.17: 82244782-82244782 on Build GRCh38
rs201284247 Chr.17: 82244858-82244858 on Build GRCh38
rs759798079 Chr.17: 82244822-82244822 on Build GRCh38
rs763350495 Chr.17: 82244852-82244853 on Build GRCh38
rs772212591 Chr.17: 82244881-82244881 on Build GRCh38
rs746234868 Chr.17: 82244697-82244697 on Build GRCh38
rs750100373 Chr.17: 82244841-82244841 on Build GRCh38
rs779652646 Chr.17: 82244866-82244866 on Build GRCh38
rs749160505 Chr.17: 82244872-82244872 on Build GRCh38
rs547614079 Chr.17: 82244749-82244749 on Build GRCh38
rs188646011 Chr.17: 82244773-82244773 on Build GRCh38
rs887225639 Chr.17: 82244722-82244722 on Build GRCh38
rs756708959 Chr.17: 82244750-82244750 on Build GRCh38
rs371742228 Chr.17: 82244731-82244731 on Build GRCh38
rs946395872 Chr.17: 82244889-82244889 on Build GRCh38
rs139129912 Chr.17: 82244704-82244704 on Build GRCh38
rs369933773 Chr.17: 82244837-82244837 on Build GRCh38
rs375995323 Chr.17: 82244733-82244733 on Build GRCh38
rs755470214 Chr.17: 82244744-82244744 on Build GRCh38
rs745459371 Chr.17: 82244758-82244758 on Build GRCh38

View Full Product Details