Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 4068866-4069113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM217A
Entrez Gene ID
222826
Gene Name
family with sequence similarity 217 member A
Gene Aliases
C6orf146
UniGene
Hs.657342
Gene Chromosome Location
Chr.6: 4068352-4087344 on Build GRCh38
Transcripts
NM_173563.2 XM_005248951.3 XM_006715025.2 XM_006715026.3 XM_011514413.2 XM_011514414.2 XM_011514415.1 XM_011514416.2 XM_011514417.1 XM_017010480.1 XM_017010481.1 XM_017010482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6659498 c.1142C>A p.S381Y Chr.6: 4069081-4069081 on Build GRCh38
1079133 c.1239A>G p.E413E Chr.6: 4068984-4068984 on Build GRCh38
3430654 c.1326G>A p.M442I Chr.6: 4068897-4068897 on Build GRCh38
6578956 c.1217C>G p.S406C Chr.6: 4069006-4069006 on Build GRCh38
5061776 c.1113G>A p.W371* Chr.6: 4069110-4069110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144365949 Chr.6: 4068990-4068990 on Build GRCh38
rs750578108 Chr.6: 4069061-4069061 on Build GRCh38
rs767241404 Chr.6: 4068992-4068992 on Build GRCh38
rs755168765 Chr.6: 4068919-4068919 on Build GRCh38
rs757866954 Chr.6: 4068889-4068889 on Build GRCh38
rs747288896 Chr.6: 4068906-4068906 on Build GRCh38
rs751888907 Chr.6: 4069050-4069050 on Build GRCh38
rs760169990 Chr.6: 4069006-4069006 on Build GRCh38
rs748178700 Chr.6: 4068943-4068943 on Build GRCh38
rs759652397 Chr.6: 4069061-4069061 on Build GRCh38
rs865950908 Chr.6: 4069041-4069041 on Build GRCh38
rs773268608 Chr.6: 4068965-4068967 on Build GRCh38
rs771697102 Chr.6: 4068932-4068932 on Build GRCh38
rs774268947 Chr.6: 4068980-4068980 on Build GRCh38
rs372639777 Chr.6: 4069046-4069046 on Build GRCh38
rs963912502 Chr.6: 4069078-4069078 on Build GRCh38
rs770820767 Chr.6: 4068958-4068958 on Build GRCh38
rs771186708 Chr.6: 4069023-4069035 on Build GRCh38
rs765663000 Chr.6: 4069007-4069007 on Build GRCh38
rs775159894 Chr.6: 4068994-4068994 on Build GRCh38
rs369483912 Chr.6: 4068965-4068965 on Build GRCh38
rs754821576 Chr.6: 4069024-4069024 on Build GRCh38
rs752778909 Chr.6: 4069062-4069062 on Build GRCh38
rs371366203 Chr.6: 4069026-4069026 on Build GRCh38
rs766777181 Chr.6: 4069036-4069036 on Build GRCh38
rs575875550 Chr.6: 4069009-4069009 on Build GRCh38
rs377007600 Chr.6: 4068983-4068983 on Build GRCh38
rs595413 Chr.6: 4068932-4068932 on Build GRCh38
rs10485172 Chr.6: 4068899-4068899 on Build GRCh38
rs545235485 Chr.6: 4068944-4068944 on Build GRCh38
rs199724850 Chr.6: 4068953-4068953 on Build GRCh38
rs755217569 Chr.6: 4069051-4069051 on Build GRCh38
rs756283867 Chr.6: 4069074-4069074 on Build GRCh38
rs776823363 Chr.6: 4069039-4069042 on Build GRCh38
rs760518162 Chr.6: 4069023-4069025 on Build GRCh38
rs780277571 Chr.6: 4068904-4068904 on Build GRCh38
rs773135122 Chr.6: 4068950-4068950 on Build GRCh38

View Full Product Details