Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102939703-102939952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42BPB
Entrez Gene ID
9578
Gene Name
CDC42 binding protein kinase beta
Gene Aliases
MRCKB
UniGene
Hs.654634
Gene Chromosome Location
Chr.14: 102932379-103057405 on Build GRCh38
Transcripts
NM_006035.3 XM_005268227.1 XM_005268228.1 XM_005268229.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1732750 c.4646G>A p.R1549H Chr.14: 102939893-102939893 on Build GRCh38
5893757 c.4619C>T p.S1540F Chr.14: 102939920-102939920 on Build GRCh38
7096298 c.4674C>T p.F1558F Chr.14: 102939865-102939865 on Build GRCh38
2025250 c.4668C>T p.F1556F Chr.14: 102939871-102939871 on Build GRCh38
6624387 c.4621G>A p.D1541N Chr.14: 102939918-102939918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779495697 Chr.14: 102939751-102939751 on Build GRCh38
rs779185138 Chr.14: 102939818-102939818 on Build GRCh38
rs758971557 Chr.14: 102939822-102939822 on Build GRCh38
rs368319342 Chr.14: 102939794-102939794 on Build GRCh38
rs775479309 Chr.14: 102939891-102939891 on Build GRCh38
rs745958701 Chr.14: 102939757-102939757 on Build GRCh38
rs762832845 Chr.14: 102939898-102939898 on Build GRCh38
rs563814555 Chr.14: 102939928-102939928 on Build GRCh38
rs759399853 Chr.14: 102939772-102939772 on Build GRCh38
rs772766652 Chr.14: 102939756-102939756 on Build GRCh38
rs770300795 Chr.14: 102939863-102939863 on Build GRCh38
rs17101081 Chr.14: 102939738-102939738 on Build GRCh38
rs772535709 Chr.14: 102939753-102939753 on Build GRCh38
rs760828996 Chr.14: 102939916-102939916 on Build GRCh38
rs775397673 Chr.14: 102939788-102939788 on Build GRCh38
rs769470267 Chr.14: 102939874-102939874 on Build GRCh38
rs371308106 Chr.14: 102939763-102939763 on Build GRCh38
rs374818759 Chr.14: 102939770-102939770 on Build GRCh38
rs753065038 Chr.14: 102939925-102939925 on Build GRCh38
rs140004721 Chr.14: 102939830-102939830 on Build GRCh38
rs1009900223 Chr.14: 102939918-102939918 on Build GRCh38
rs774370821 Chr.14: 102939911-102939911 on Build GRCh38
rs780932163 Chr.14: 102939870-102939870 on Build GRCh38
rs544013491 Chr.14: 102939915-102939915 on Build GRCh38
rs368182073 Chr.14: 102939783-102939783 on Build GRCh38
rs750593843 Chr.14: 102939802-102939802 on Build GRCh38
rs768715462 Chr.14: 102939900-102939900 on Build GRCh38
rs745550984 Chr.14: 102939871-102939871 on Build GRCh38
rs375217140 Chr.14: 102939732-102939732 on Build GRCh38
rs763037873 Chr.14: 102939792-102939792 on Build GRCh38
rs752024361 Chr.14: 102939929-102939929 on Build GRCh38
rs759726532 Chr.14: 102939920-102939920 on Build GRCh38
rs748499505 Chr.14: 102939821-102939821 on Build GRCh38
rs373755648 Chr.14: 102939875-102939875 on Build GRCh38
rs60475222 Chr.14: 102939832-102939832 on Build GRCh38
rs200207520 Chr.14: 102939752-102939752 on Build GRCh38
rs755400950 Chr.14: 102939817-102939817 on Build GRCh38
rs766329158 Chr.14: 102939917-102939917 on Build GRCh38
rs774624337 Chr.14: 102939742-102939742 on Build GRCh38
rs188937393 Chr.14: 102939811-102939811 on Build GRCh38
rs373428218 Chr.14: 102939812-102939812 on Build GRCh38
rs745708875 Chr.14: 102939779-102939780 on Build GRCh38
rs758705817 Chr.14: 102939926-102939926 on Build GRCh38
rs1019328882 Chr.14: 102939886-102939886 on Build GRCh38
rs765442114 Chr.14: 102939923-102939923 on Build GRCh38
rs139297022 Chr.14: 102939817-102939818 on Build GRCh38
rs776437116 Chr.14: 102939922-102939922 on Build GRCh38
rs202209179 Chr.14: 102939933-102939933 on Build GRCh38
rs753957301 Chr.14: 102939816-102939816 on Build GRCh38
rs753922027 Chr.14: 102939919-102939919 on Build GRCh38

View Full Product Details