Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73450788-73451049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAZ1B
Entrez Gene ID
9031
Gene Name
bromodomain adjacent to zinc finger domain 1B
Gene Aliases
WBSCR10, WBSCR9, WSTF
UniGene
Hs.743372
Gene Chromosome Location
Chr.7: 73440398-73522298 on Build GRCh38
Transcripts
NM_032408.3 XM_017012773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6047693 c.3500T>C p.V1167A Chr.7: 73450927-73450927 on Build GRCh38
3674973 c.3470G>T p.R1157L Chr.7: 73450957-73450957 on Build GRCh38
747178 c.3547G>T p.E1183* Chr.7: 73450880-73450880 on Build GRCh38
1451969 c.3559T>C p.C1187R Chr.7: 73450868-73450868 on Build GRCh38
5873033 c.3542_3543CC>TT p.S1181F Chr.7: 73450884-73450885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776700957 Chr.7: 73450930-73450930 on Build GRCh38
rs896353691 Chr.7: 73450945-73450945 on Build GRCh38
rs782726863 Chr.7: 73451006-73451006 on Build GRCh38
rs1023530227 Chr.7: 73450985-73450985 on Build GRCh38
rs143371789 Chr.7: 73450881-73450881 on Build GRCh38
rs368624518 Chr.7: 73450903-73450903 on Build GRCh38
rs782106356 Chr.7: 73451004-73451004 on Build GRCh38
rs148299208 Chr.7: 73450884-73450884 on Build GRCh38
rs782309014 Chr.7: 73450956-73450956 on Build GRCh38
rs782545578 Chr.7: 73450926-73450926 on Build GRCh38
rs781989876 Chr.7: 73450989-73450989 on Build GRCh38
rs782051515 Chr.7: 73450826-73450826 on Build GRCh38
rs201633373 Chr.7: 73450911-73450911 on Build GRCh38
rs782035004 Chr.7: 73450969-73450969 on Build GRCh38
rs782344823 Chr.7: 73450818-73450818 on Build GRCh38
rs781783517 Chr.7: 73450923-73450923 on Build GRCh38
rs182673668 Chr.7: 73451018-73451018 on Build GRCh38
rs782811497 Chr.7: 73450855-73450855 on Build GRCh38
rs200721268 Chr.7: 73450993-73450993 on Build GRCh38
rs782024430 Chr.7: 73450869-73450869 on Build GRCh38
rs374865536 Chr.7: 73450974-73450974 on Build GRCh38
rs781937255 Chr.7: 73450821-73450821 on Build GRCh38
rs782262083 Chr.7: 73450944-73450944 on Build GRCh38
rs768610254 Chr.7: 73450888-73450888 on Build GRCh38
rs782502569 Chr.7: 73450947-73450947 on Build GRCh38
rs1013438793 Chr.7: 73450958-73450958 on Build GRCh38
rs1025423767 Chr.7: 73450934-73450934 on Build GRCh38
rs372715830 Chr.7: 73450950-73450950 on Build GRCh38
rs782769772 Chr.7: 73450883-73450883 on Build GRCh38
rs558448410 Chr.7: 73450935-73450935 on Build GRCh38
rs782397109 Chr.7: 73450977-73450977 on Build GRCh38

View Full Product Details