Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30836597-30836866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM188B
Entrez Gene ID
84182
Gene Name
family with sequence similarity 188 member B
Gene Aliases
C7orf67
UniGene
Hs.660192
Gene Chromosome Location
Chr.7: 30771417-30892387 on Build GRCh38
Transcripts
NM_032222.2 NR_037598.1

Gene Symbol
INMT-FAM188B
Entrez Gene ID
100526825
Gene Name
INMT-FAM188B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.660192
Gene Chromosome Location
Chr.7: 30752135-30892387 on Build GRCh38
Transcripts
NM_032222.2 NR_037598.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
421476 c.1229C>T p.S410L Chr.7: 30836754-30836754 on Build GRCh38
4568536 c.1198T>C p.L400L Chr.7: 30836723-30836723 on Build GRCh38
4682808 c.1175C>T p.S392L Chr.7: 30836700-30836700 on Build GRCh38
244381 c.1219A>G p.I407V Chr.7: 30836744-30836744 on Build GRCh38
3028457 c.1229C>G p.S410* Chr.7: 30836754-30836754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766167580 Chr.7: 30836725-30836725 on Build GRCh38
rs778773285 Chr.7: 30836623-30836623 on Build GRCh38
rs914560916 Chr.7: 30836796-30836796 on Build GRCh38
rs187366942 Chr.7: 30836700-30836700 on Build GRCh38
rs566476872 Chr.7: 30836823-30836823 on Build GRCh38
rs538935656 Chr.7: 30836832-30836832 on Build GRCh38
rs772351858 Chr.7: 30836806-30836806 on Build GRCh38
rs752372079 Chr.7: 30836745-30836745 on Build GRCh38
rs948712099 Chr.7: 30836801-30836801 on Build GRCh38
rs762153582 Chr.7: 30836804-30836804 on Build GRCh38
rs201335083 Chr.7: 30836754-30836754 on Build GRCh38
rs746420213 Chr.7: 30836632-30836632 on Build GRCh38
rs774522399 Chr.7: 30836794-30836794 on Build GRCh38
rs369212440 Chr.7: 30836708-30836708 on Build GRCh38
rs746162457 Chr.7: 30836758-30836758 on Build GRCh38
rs758007151 Chr.7: 30836753-30836753 on Build GRCh38
rs764443009 Chr.7: 30836653-30836653 on Build GRCh38
rs770391410 Chr.7: 30836766-30836766 on Build GRCh38
rs771418950 Chr.7: 30836651-30836651 on Build GRCh38
rs747671942 Chr.7: 30836645-30836645 on Build GRCh38
rs753701081 Chr.7: 30836815-30836815 on Build GRCh38
rs200634054 Chr.7: 30836750-30836750 on Build GRCh38
rs533987127 Chr.7: 30836625-30836625 on Build GRCh38
rs749424254 Chr.7: 30836777-30836777 on Build GRCh38
rs776933145 Chr.7: 30836630-30836630 on Build GRCh38
rs374910173 Chr.7: 30836744-30836744 on Build GRCh38
rs557402928 Chr.7: 30836841-30836841 on Build GRCh38
rs191702079 Chr.7: 30836787-30836787 on Build GRCh38
rs10258097 Chr.7: 30836816-30836816 on Build GRCh38
rs754417879 Chr.7: 30836736-30836736 on Build GRCh38
rs373120157 Chr.7: 30836813-30836813 on Build GRCh38
rs780418809 Chr.7: 30836771-30836771 on Build GRCh38
rs375867200 Chr.7: 30836626-30836626 on Build GRCh38
rs775653982 Chr.7: 30836637-30836637 on Build GRCh38
rs905893541 Chr.7: 30836682-30836682 on Build GRCh38
rs368634456 Chr.7: 30836649-30836649 on Build GRCh38
rs773069635 Chr.7: 30836808-30836808 on Build GRCh38
rs530846398 Chr.7: 30836651-30836651 on Build GRCh38
rs141888334 Chr.7: 30836701-30836701 on Build GRCh38
rs371516368 Chr.7: 30836727-30836727 on Build GRCh38
rs760664202 Chr.7: 30836710-30836710 on Build GRCh38
rs968839025 Chr.7: 30836822-30836822 on Build GRCh38
rs771266975 Chr.7: 30836628-30836628 on Build GRCh38
rs774681733 Chr.7: 30836689-30836689 on Build GRCh38
rs995632383 Chr.7: 30836643-30836643 on Build GRCh38
rs766383337 Chr.7: 30836814-30836814 on Build GRCh38

View Full Product Details