Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 961947-962184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKQ
Entrez Gene ID
1609
Gene Name
diacylglycerol kinase theta
Gene Aliases
DAGK, DAGK4, DAGK7
UniGene
Hs.584858
Gene Chromosome Location
Chr.4: 958884-973560 on Build GRCh38
Transcripts
NM_001347.3 XM_011513411.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3607125 c.2305T>C p.F769L Chr.4: 961992-961992 on Build GRCh38
448354 c.2262G>C p.L754L Chr.4: 962035-962035 on Build GRCh38
5459128 c.2256G>A p.A752A Chr.4: 962041-962041 on Build GRCh38
1158329 c.2253C>G p.D751E Chr.4: 962044-962044 on Build GRCh38
5975281 c.2218G>A p.V740M Chr.4: 962079-962079 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758794505 Chr.4: 962096-962096 on Build GRCh38
rs994406701 Chr.4: 962157-962157 on Build GRCh38
rs766313922 Chr.4: 961998-961998 on Build GRCh38
rs556260739 Chr.4: 962119-962119 on Build GRCh38
rs987350759 Chr.4: 962042-962042 on Build GRCh38
rs965505320 Chr.4: 962027-962027 on Build GRCh38
rs746394457 Chr.4: 961968-961968 on Build GRCh38
rs765157641 Chr.4: 961982-961982 on Build GRCh38
rs973243511 Chr.4: 962019-962019 on Build GRCh38
rs548721273 Chr.4: 962041-962041 on Build GRCh38
rs150502750 Chr.4: 962053-962053 on Build GRCh38
rs928117506 Chr.4: 962083-962083 on Build GRCh38
rs759984244 Chr.4: 962091-962092 on Build GRCh38
rs1014676117 Chr.4: 962011-962011 on Build GRCh38
rs773577653 Chr.4: 962121-962121 on Build GRCh38
rs35719180 Chr.4: 962044-962044 on Build GRCh38
rs116777977 Chr.4: 962135-962135 on Build GRCh38
rs536470681 Chr.4: 961976-961976 on Build GRCh38
rs376888708 Chr.4: 961970-961970 on Build GRCh38
rs906233245 Chr.4: 962099-962099 on Build GRCh38
rs767047009 Chr.4: 962088-962088 on Build GRCh38
rs770195369 Chr.4: 962118-962118 on Build GRCh38
rs187639231 Chr.4: 962117-962117 on Build GRCh38
rs759345186 Chr.4: 962080-962080 on Build GRCh38
rs534501480 Chr.4: 962059-962059 on Build GRCh38
rs936055431 Chr.4: 962058-962058 on Build GRCh38
rs747571257 Chr.4: 962100-962100 on Build GRCh38
rs375662322 Chr.4: 962094-962094 on Build GRCh38
rs115438176 Chr.4: 961971-961971 on Build GRCh38
rs754527129 Chr.4: 962018-962018 on Build GRCh38
rs373161518 Chr.4: 962047-962047 on Build GRCh38
rs766944584 Chr.4: 962020-962020 on Build GRCh38
rs541669195 Chr.4: 962137-962137 on Build GRCh38
rs901042609 Chr.4: 962156-962156 on Build GRCh38
rs751084332 Chr.4: 962013-962013 on Build GRCh38
rs558554660 Chr.4: 962091-962091 on Build GRCh38
rs370196598 Chr.4: 962095-962095 on Build GRCh38
rs770785648 Chr.4: 962128-962128 on Build GRCh38
rs549888127 Chr.4: 961966-961966 on Build GRCh38
rs372753778 Chr.4: 962113-962113 on Build GRCh38
rs749416760 Chr.4: 962126-962126 on Build GRCh38
rs762061695 Chr.4: 961977-961977 on Build GRCh38
rs770122049 Chr.4: 962063-962063 on Build GRCh38
rs374013481 Chr.4: 962079-962079 on Build GRCh38
rs750197208 Chr.4: 961984-961984 on Build GRCh38
rs935488431 Chr.4: 962092-962092 on Build GRCh38
rs893864866 Chr.4: 962102-962102 on Build GRCh38
rs776814998 Chr.4: 962054-962054 on Build GRCh38
rs781458641 Chr.4: 962115-962115 on Build GRCh38
rs369895580 Chr.4: 962073-962073 on Build GRCh38
rs199709114 Chr.4: 962068-962068 on Build GRCh38
rs137972534 Chr.4: 962043-962043 on Build GRCh38
rs147376704 Chr.4: 962039-962039 on Build GRCh38
rs773979205 Chr.4: 962130-962130 on Build GRCh38
rs762851857 Chr.4: 961988-961988 on Build GRCh38
rs768139104 Chr.4: 962134-962135 on Build GRCh38
rs768819107 Chr.4: 961973-961973 on Build GRCh38
rs755713610 Chr.4: 962026-962026 on Build GRCh38
rs773185079 Chr.4: 962065-962065 on Build GRCh38
rs747760873 Chr.4: 962023-962023 on Build GRCh38
rs747418602 Chr.4: 962051-962051 on Build GRCh38

View Full Product Details