Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50079857-50080106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC1
Entrez Gene ID
41
Gene Name
acid sensing ion channel subunit 1
Gene Aliases
ACCN2, ASIC, BNaC2
UniGene
Hs.274361
Gene Chromosome Location
Chr.12: 50057637-50083622 on Build GRCh38
Transcripts
NM_001095.3 NM_001256830.1 NM_020039.3 NR_046389.1 XM_011538350.1 XM_011538351.2 XM_011538352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1188489 c.1201A>G p.I401V Chr.12: 50080051-50080051 on Build GRCh38
1726081 c.1115A>G p.Y372C Chr.12: 50079965-50079965 on Build GRCh38
3461791 c.1090G>A p.E364K Chr.12: 50079940-50079940 on Build GRCh38
1737136 c.1112G>A p.R371H Chr.12: 50079962-50079962 on Build GRCh38
7148694 c.1176G>T p.K392N Chr.12: 50080026-50080026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773760004 Chr.12: 50079909-50079909 on Build GRCh38
rs200653479 Chr.12: 50079922-50079922 on Build GRCh38
rs758091036 Chr.12: 50079966-50079966 on Build GRCh38
rs2307080 Chr.12: 50079972-50079972 on Build GRCh38
rs404751 Chr.12: 50079877-50079877 on Build GRCh38
rs766860378 Chr.12: 50079955-50079955 on Build GRCh38
rs201684162 Chr.12: 50080044-50080044 on Build GRCh38
rs774238627 Chr.12: 50079891-50079891 on Build GRCh38
rs752105040 Chr.12: 50079962-50079962 on Build GRCh38
rs751416549 Chr.12: 50079991-50079991 on Build GRCh38
rs574384213 Chr.12: 50079885-50079885 on Build GRCh38
rs959943518 Chr.12: 50079931-50079931 on Build GRCh38
rs377261511 Chr.12: 50079888-50079888 on Build GRCh38
rs764823218 Chr.12: 50079961-50079961 on Build GRCh38
rs779080982 Chr.12: 50080079-50080079 on Build GRCh38
rs754372339 Chr.12: 50079959-50079959 on Build GRCh38
rs757044510 Chr.12: 50079993-50079993 on Build GRCh38
rs544797255 Chr.12: 50080062-50080062 on Build GRCh38
rs199923697 Chr.12: 50079934-50079934 on Build GRCh38
rs537535975 Chr.12: 50079899-50079899 on Build GRCh38
rs575174823 Chr.12: 50079898-50079898 on Build GRCh38
rs748216409 Chr.12: 50080084-50080084 on Build GRCh38
rs143293039 Chr.12: 50079960-50079960 on Build GRCh38
rs201655846 Chr.12: 50080005-50080005 on Build GRCh38
rs745831832 Chr.12: 50080050-50080050 on Build GRCh38

View Full Product Details