Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38473416-38473681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM98B
Entrez Gene ID
283742
Gene Name
family with sequence similarity 98 member B
Gene Aliases
-
UniGene
Hs.625289
Gene Chromosome Location
Chr.15: 38454127-38487710 on Build GRCh38
Transcripts
NM_173611.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3500701 c.602C>T p.S201L Chr.15: 38473575-38473575 on Build GRCh38
960987 c.572G>A p.G191E Chr.15: 38473545-38473545 on Build GRCh38
7367565 c.552G>T p.K184N Chr.15: 38473525-38473525 on Build GRCh38
960988 c.572G>A p.G191E Chr.15: 38473545-38473545 on Build GRCh38
4499732 c.548C>T p.S183L Chr.15: 38473521-38473521 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770147755 Chr.15: 38473458-38473458 on Build GRCh38
rs569984690 Chr.15: 38473497-38473497 on Build GRCh38
rs761365550 Chr.15: 38473521-38473521 on Build GRCh38
rs749797212 Chr.15: 38473485-38473485 on Build GRCh38
rs201011129 Chr.15: 38473515-38473515 on Build GRCh38
rs766223410 Chr.15: 38473585-38473585 on Build GRCh38
rs537349939 Chr.15: 38473498-38473498 on Build GRCh38
rs1048341961 Chr.15: 38473511-38473511 on Build GRCh38
rs755206687 Chr.15: 38473601-38473601 on Build GRCh38
rs771321555 Chr.15: 38473491-38473491 on Build GRCh38
rs748286460 Chr.15: 38473455-38473455 on Build GRCh38
rs139669477 Chr.15: 38473505-38473505 on Build GRCh38
rs767266136 Chr.15: 38473622-38473622 on Build GRCh38
rs187994626 Chr.15: 38473460-38473460 on Build GRCh38
rs764696390 Chr.15: 38473550-38473550 on Build GRCh38
rs370979515 Chr.15: 38473636-38473636 on Build GRCh38
rs751401448 Chr.15: 38473595-38473595 on Build GRCh38
rs780016334 Chr.15: 38473616-38473616 on Build GRCh38
rs371947818 Chr.15: 38473602-38473602 on Build GRCh38
rs751280530 Chr.15: 38473539-38473540 on Build GRCh38
rs918498504 Chr.15: 38473451-38473451 on Build GRCh38
rs769687413 Chr.15: 38473584-38473584 on Build GRCh38
rs984551599 Chr.15: 38473640-38473640 on Build GRCh38
rs887748802 Chr.15: 38473534-38473534 on Build GRCh38
rs992688210 Chr.15: 38473445-38473445 on Build GRCh38
rs752908526 Chr.15: 38473608-38473608 on Build GRCh38
rs778318368 Chr.15: 38473484-38473484 on Build GRCh38
rs573885858 Chr.15: 38473563-38473563 on Build GRCh38
rs754726883 Chr.15: 38473549-38473550 on Build GRCh38

View Full Product Details