Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 85328585-85328816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA1
Entrez Gene ID
759
Gene Name
carbonic anhydrase 1
Gene Aliases
CA-I, CAB, Car1, HEL-S-11
UniGene
Hs.23118
Gene Chromosome Location
Chr.8: 85328229-85378154 on Build GRCh38
Transcripts
NM_001128829.3 NM_001128830.3 NM_001128831.3 NM_001164830.1 NM_001291967.1 NM_001291968.1 NM_001738.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1673985 c.739C>A p.R247S Chr.8: 85328607-85328607 on Build GRCh38
486715 c.705A>G p.E235E Chr.8: 85328641-85328641 on Build GRCh38
3651392 c.681C>T p.F227F Chr.8: 85328665-85328665 on Build GRCh38
6258116 c.746C>A p.T249N Chr.8: 85328600-85328600 on Build GRCh38
1102101 c.686G>T p.S229I Chr.8: 85328660-85328660 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943445568 Chr.8: 85328614-85328614 on Build GRCh38
rs764614596 Chr.8: 85328698-85328698 on Build GRCh38
rs756681346 Chr.8: 85328608-85328608 on Build GRCh38
rs771935078 Chr.8: 85328630-85328630 on Build GRCh38
rs757305470 Chr.8: 85328626-85328626 on Build GRCh38
rs761150220 Chr.8: 85328695-85328695 on Build GRCh38
rs952024120 Chr.8: 85328751-85328751 on Build GRCh38
rs769195145 Chr.8: 85328664-85328664 on Build GRCh38
rs896234801 Chr.8: 85328732-85328732 on Build GRCh38
rs754627939 Chr.8: 85328652-85328652 on Build GRCh38
rs772797236 Chr.8: 85328665-85328665 on Build GRCh38
rs1001914259 Chr.8: 85328726-85328726 on Build GRCh38
rs767885397 Chr.8: 85328607-85328607 on Build GRCh38
rs199514200 Chr.8: 85328656-85328656 on Build GRCh38
rs762656239 Chr.8: 85328670-85328670 on Build GRCh38
rs753319731 Chr.8: 85328619-85328619 on Build GRCh38
rs777685360 Chr.8: 85328621-85328621 on Build GRCh38
rs78898210 Chr.8: 85328622-85328622 on Build GRCh38
rs753101577 Chr.8: 85328687-85328687 on Build GRCh38
rs371314891 Chr.8: 85328627-85328627 on Build GRCh38
rs139959399 Chr.8: 85328663-85328663 on Build GRCh38
rs757293070 Chr.8: 85328714-85328714 on Build GRCh38
rs1044583934 Chr.8: 85328667-85328667 on Build GRCh38
rs146043953 Chr.8: 85328632-85328632 on Build GRCh38
rs914852944 Chr.8: 85328658-85328658 on Build GRCh38
rs1014413295 Chr.8: 85328634-85328634 on Build GRCh38
rs141945543 Chr.8: 85328631-85328631 on Build GRCh38
rs955184362 Chr.8: 85328761-85328761 on Build GRCh38
rs1026378775 Chr.8: 85328734-85328734 on Build GRCh38
rs993214586 Chr.8: 85328733-85328733 on Build GRCh38
rs77332660 Chr.8: 85328722-85328722 on Build GRCh38
rs544308306 Chr.8: 85328729-85328729 on Build GRCh38
rs758755442 Chr.8: 85328728-85328728 on Build GRCh38
rs74427886 Chr.8: 85328752-85328752 on Build GRCh38
rs752181935 Chr.8: 85328672-85328672 on Build GRCh38
rs1020421101 Chr.8: 85328755-85328755 on Build GRCh38
rs778871775 Chr.8: 85328717-85328717 on Build GRCh38
rs121909578 Chr.8: 85328606-85328606 on Build GRCh38
rs574224726 Chr.8: 85328760-85328760 on Build GRCh38
rs754330305 Chr.8: 85328704-85328704 on Build GRCh38
rs777641626 Chr.8: 85328682-85328696 on Build GRCh38
rs543234924 Chr.8: 85328784-85328784 on Build GRCh38
rs560124122 Chr.8: 85328785-85328785 on Build GRCh38
rs768675511 Chr.8: 85328644-85328644 on Build GRCh38

View Full Product Details