Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 91962638-91962879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRDT
Entrez Gene ID
676
Gene Name
bromodomain testis associated
Gene Aliases
BRD6, CT9
UniGene
Hs.482520
Gene Chromosome Location
Chr.1: 91949371-92014428 on Build GRCh38
Transcripts
NM_001242805.2 NM_001242806.2 NM_001242807.2 NM_001242808.2 NM_001242810.2 NM_001726.4 NM_207189.3 XM_006710853.3 XM_006710854.3 XM_006710855.3 XM_006710856.3 XM_006710857.3 XM_011542032.2 XM_011542033.2 XM_011542034.2 XM_011542035.2 XM_011542036.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6507500 c.82T>A p.L28M Chr.1: 91962836-91962836 on Build GRCh38
3866239 c.94C>A p.L32I Chr.1: 91962848-91962848 on Build GRCh38
6305963 c.73A>T p.N25Y Chr.1: 91962827-91962827 on Build GRCh38
4889105 c.116T>C p.V39A Chr.1: 91962870-91962870 on Build GRCh38
389808 c.17G>A p.R6Q Chr.1: 91962771-91962771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045296097 Chr.1: 91962675-91962675 on Build GRCh38
rs1001582009 Chr.1: 91962687-91962687 on Build GRCh38
rs758284638 Chr.1: 91962750-91962755 on Build GRCh38
rs753146796 Chr.1: 91962844-91962844 on Build GRCh38
rs553069454 Chr.1: 91962677-91962677 on Build GRCh38
rs779686999 Chr.1: 91962781-91962783 on Build GRCh38
rs779401099 Chr.1: 91962843-91962843 on Build GRCh38
rs771510328 Chr.1: 91962772-91962772 on Build GRCh38
rs548301739 Chr.1: 91962821-91962821 on Build GRCh38
rs994701932 Chr.1: 91962674-91962674 on Build GRCh38
rs769873755 Chr.1: 91962795-91962795 on Build GRCh38
rs767875806 Chr.1: 91962736-91962736 on Build GRCh38
rs777405645 Chr.1: 91962779-91962779 on Build GRCh38
rs895702373 Chr.1: 91962701-91962701 on Build GRCh38
rs377425843 Chr.1: 91962738-91962738 on Build GRCh38
rs200800858 Chr.1: 91962722-91962722 on Build GRCh38
rs754180862 Chr.1: 91962838-91962838 on Build GRCh38
rs745497883 Chr.1: 91962722-91962727 on Build GRCh38
rs759352809 Chr.1: 91962792-91962792 on Build GRCh38
rs759449186 Chr.1: 91962715-91962715 on Build GRCh38
rs750499319 Chr.1: 91962830-91962830 on Build GRCh38
rs751729226 Chr.1: 91962719-91962719 on Build GRCh38
rs779348247 Chr.1: 91962770-91962770 on Build GRCh38
rs754221182 Chr.1: 91962760-91962760 on Build GRCh38
rs770603753 Chr.1: 91962788-91962788 on Build GRCh38
rs765198207 Chr.1: 91962716-91962716 on Build GRCh38
rs746748695 Chr.1: 91962794-91962796 on Build GRCh38
rs775526570 Chr.1: 91962797-91962797 on Build GRCh38
rs55806733 Chr.1: 91962759-91962759 on Build GRCh38
rs760869013 Chr.1: 91962831-91962831 on Build GRCh38
rs1020559427 Chr.1: 91962733-91962733 on Build GRCh38
rs370161455 Chr.1: 91962703-91962703 on Build GRCh38
rs573028112 Chr.1: 91962801-91962801 on Build GRCh38
rs199720664 Chr.1: 91962765-91962765 on Build GRCh38
rs746705309 Chr.1: 91962782-91962782 on Build GRCh38
rs1057414384 Chr.1: 91962699-91962699 on Build GRCh38
rs756508455 Chr.1: 91962757-91962757 on Build GRCh38
rs138274836 Chr.1: 91962847-91962847 on Build GRCh38
rs535480538 Chr.1: 91962711-91962711 on Build GRCh38
rs755333661 Chr.1: 91962839-91962839 on Build GRCh38
rs767644399 Chr.1: 91962820-91962820 on Build GRCh38
rs150325383 Chr.1: 91962685-91962685 on Build GRCh38
rs953489700 Chr.1: 91962796-91962796 on Build GRCh38
rs56273490 Chr.1: 91962771-91962771 on Build GRCh38
rs776339706 Chr.1: 91962790-91962790 on Build GRCh38
rs750618078 Chr.1: 91962740-91962740 on Build GRCh38
rs1035697556 Chr.1: 91962702-91962702 on Build GRCh38

View Full Product Details