Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203180441-203180680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHI3L1
Entrez Gene ID
1116
Gene Name
chitinase 3 like 1
Gene Aliases
ASRT7, CGP-39, GP-39, GP39, HC-gp39, HCGP-3P, YKL-40, YKL40, YYL-40, hCGP-39
UniGene
Hs.382202
Gene Chromosome Location
Chr.1: 203178931-203186794 on Build GRCh38
Transcripts
NM_001276.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5025490 c.799C>G p.L267V Chr.1: 203180565-203180565 on Build GRCh38
425047 c.828C>T p.A276A Chr.1: 203180536-203180536 on Build GRCh38
234541 c.783C>T p.F261F Chr.1: 203180581-203180581 on Build GRCh38
3360578 c.837A>C p.S279S Chr.1: 203180527-203180527 on Build GRCh38
6582509 c.779_780insAC p.F261fs*67 Chr.1: 203180584-203180585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753309927 Chr.1: 203180611-203180611 on Build GRCh38
rs764432178 Chr.1: 203180469-203180469 on Build GRCh38
rs529903795 Chr.1: 203180609-203180609 on Build GRCh38
rs868399905 Chr.1: 203180589-203180589 on Build GRCh38
rs76446715 Chr.1: 203180507-203180507 on Build GRCh38
rs779202083 Chr.1: 203180661-203180661 on Build GRCh38
rs763367362 Chr.1: 203180478-203180479 on Build GRCh38
rs200476432 Chr.1: 203180547-203180547 on Build GRCh38
rs778571121 Chr.1: 203180521-203180521 on Build GRCh38
rs760473998 Chr.1: 203180643-203180643 on Build GRCh38
rs199579588 Chr.1: 203180499-203180499 on Build GRCh38
rs775586820 Chr.1: 203180552-203180552 on Build GRCh38
rs754579093 Chr.1: 203180519-203180519 on Build GRCh38
rs138989209 Chr.1: 203180508-203180508 on Build GRCh38
rs764778624 Chr.1: 203180620-203180620 on Build GRCh38
rs749409296 Chr.1: 203180572-203180572 on Build GRCh38
rs144493623 Chr.1: 203180641-203180641 on Build GRCh38
rs780960257 Chr.1: 203180656-203180656 on Build GRCh38
rs777563441 Chr.1: 203180634-203180634 on Build GRCh38
rs577114464 Chr.1: 203180482-203180482 on Build GRCh38
rs896357573 Chr.1: 203180660-203180660 on Build GRCh38
rs772869145 Chr.1: 203180631-203180631 on Build GRCh38
rs766707810 Chr.1: 203180626-203180637 on Build GRCh38
rs767921141 Chr.1: 203180591-203180591 on Build GRCh38
rs777493129 Chr.1: 203180538-203180538 on Build GRCh38
rs376330429 Chr.1: 203180474-203180474 on Build GRCh38
rs922808011 Chr.1: 203180544-203180544 on Build GRCh38
rs757178196 Chr.1: 203180648-203180648 on Build GRCh38
rs750929926 Chr.1: 203180501-203180501 on Build GRCh38
rs761621393 Chr.1: 203180490-203180490 on Build GRCh38
rs150868636 Chr.1: 203180581-203180581 on Build GRCh38
rs562001678 Chr.1: 203180594-203180594 on Build GRCh38
rs200949867 Chr.1: 203180580-203180580 on Build GRCh38
rs752337033 Chr.1: 203180623-203180623 on Build GRCh38
rs968078375 Chr.1: 203180481-203180481 on Build GRCh38
rs887208330 Chr.1: 203180582-203180582 on Build GRCh38
rs377356528 Chr.1: 203180659-203180659 on Build GRCh38
rs758170643 Chr.1: 203180537-203180537 on Build GRCh38
rs774893197 Chr.1: 203180637-203180637 on Build GRCh38
rs759020454 Chr.1: 203180617-203180617 on Build GRCh38
rs370805361 Chr.1: 203180522-203180522 on Build GRCh38
rs955305477 Chr.1: 203180585-203180585 on Build GRCh38
rs745850378 Chr.1: 203180657-203180657 on Build GRCh38
rs1034840968 Chr.1: 203180647-203180647 on Build GRCh38

View Full Product Details