Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31900992-31901251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2
Entrez Gene ID
717
Gene Name
complement component 2
Gene Aliases
ARMD14, CO2
UniGene
Hs.408903
Gene Chromosome Location
Chr.6: 31897785-31945674 on Build GRCh38
Transcripts
NM_001178063.2 NM_001282457.1 NM_181842.2 XM_011514383.2

Gene Symbol
ZBTB12
Entrez Gene ID
221527
Gene Name
zinc finger and BTB domain containing 12
Gene Aliases
Bat9, C6orf46, D6S59E, G10, NG35
UniGene
Hs.234027
Gene Chromosome Location
Chr.6: 31899194-31902331 on Build GRCh38
Transcripts
NM_001178063.2 NM_001282457.1 NM_181842.2 XM_011514383.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5818737 c.166C>A p.P56T Chr.6: 31901140-31901140 on Build GRCh38
1487617 c.301G>T p.V101F Chr.6: 31901005-31901005 on Build GRCh38
5782946 c.188T>C p.L63P Chr.6: 31901118-31901118 on Build GRCh38
4742072 c.273C>T p.G91G Chr.6: 31901033-31901033 on Build GRCh38
1443403 c.117C>T p.A39A Chr.6: 31901189-31901189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779376110 Chr.6: 31901204-31901204 on Build GRCh38
rs760109860 Chr.6: 31901135-31901135 on Build GRCh38
rs761572069 Chr.6: 31901084-31901084 on Build GRCh38
rs755332433 Chr.6: 31901192-31901192 on Build GRCh38
rs778141187 Chr.6: 31901226-31901226 on Build GRCh38
rs748362018 Chr.6: 31901036-31901036 on Build GRCh38
rs145104876 Chr.6: 31901215-31901215 on Build GRCh38
rs766883433 Chr.6: 31901138-31901138 on Build GRCh38
rs377556449 Chr.6: 31901060-31901060 on Build GRCh38
rs755313282 Chr.6: 31901033-31901033 on Build GRCh38
rs767142872 Chr.6: 31901105-31901105 on Build GRCh38
rs758708801 Chr.6: 31901222-31901222 on Build GRCh38
rs376539476 Chr.6: 31901051-31901051 on Build GRCh38
rs778051360 Chr.6: 31901045-31901045 on Build GRCh38
rs772154668 Chr.6: 31901043-31901043 on Build GRCh38
rs779027276 Chr.6: 31901034-31901034 on Build GRCh38
rs762675725 Chr.6: 31901063-31901063 on Build GRCh38
rs866429455 Chr.6: 31901101-31901101 on Build GRCh38
rs148838488 Chr.6: 31901066-31901066 on Build GRCh38
rs1051830114 Chr.6: 31901147-31901147 on Build GRCh38
rs773982900 Chr.6: 31901071-31901071 on Build GRCh38
rs747076075 Chr.6: 31901048-31901048 on Build GRCh38
rs749656785 Chr.6: 31901018-31901018 on Build GRCh38
rs750001934 Chr.6: 31901120-31901120 on Build GRCh38
rs866962134 Chr.6: 31901195-31901195 on Build GRCh38
rs9394078 Chr.6: 31901161-31901161 on Build GRCh38

View Full Product Details