Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152720012-152720277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf68
Entrez Gene ID
100129271
Gene Name
chromosome 1 open reading frame 68
Gene Aliases
LEP7, XP32
UniGene
Hs.601077
Gene Chromosome Location
Chr.1: 152719522-152720429 on Build GRCh38
Transcripts
NM_001024679.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3771456 c.610T>C p.S204P Chr.1: 152720131-152720131 on Build GRCh38
5689817 c.622G>C p.G208R Chr.1: 152720143-152720143 on Build GRCh38
3705001 c.744C>T p.C248C Chr.1: 152720265-152720265 on Build GRCh38
3750362 c.515G>A p.C172Y Chr.1: 152720036-152720036 on Build GRCh38
3979824 c.620C>G p.S207C Chr.1: 152720141-152720141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369426180 Chr.1: 152720132-152720132 on Build GRCh38
rs144355409 Chr.1: 152720221-152720221 on Build GRCh38
rs368436282 Chr.1: 152720039-152720039 on Build GRCh38
rs567419709 Chr.1: 152720075-152720075 on Build GRCh38
rs1045634693 Chr.1: 152720195-152720195 on Build GRCh38
rs750131088 Chr.1: 152720137-152720137 on Build GRCh38
rs571448810 Chr.1: 152720048-152720048 on Build GRCh38
rs867607238 Chr.1: 152720133-152720133 on Build GRCh38
rs59194678 Chr.1: 152720038-152720038 on Build GRCh38
rs553777962 Chr.1: 152720074-152720074 on Build GRCh38
rs533918314 Chr.1: 152720058-152720058 on Build GRCh38
rs781353683 Chr.1: 152720244-152720244 on Build GRCh38
rs770154522 Chr.1: 152720043-152720043 on Build GRCh38
rs372420974 Chr.1: 152720087-152720087 on Build GRCh38
rs771012801 Chr.1: 152720056-152720056 on Build GRCh38
rs576190431 Chr.1: 152720152-152720152 on Build GRCh38
rs971844085 Chr.1: 152720180-152720180 on Build GRCh38
rs41268474 Chr.1: 152720036-152720036 on Build GRCh38
rs757822185 Chr.1: 152720165-152720165 on Build GRCh38
rs944682 Chr.1: 152720112-152720112 on Build GRCh38
rs760632549 Chr.1: 152720055-152720055 on Build GRCh38
rs191594967 Chr.1: 152720047-152720047 on Build GRCh38
rs781462773 Chr.1: 152720200-152720200 on Build GRCh38
rs369256078 Chr.1: 152720231-152720231 on Build GRCh38
rs908373091 Chr.1: 152720031-152720031 on Build GRCh38
rs755530821 Chr.1: 152720233-152720233 on Build GRCh38

View Full Product Details