Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101611198-101611463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW4
Entrez Gene ID
6468
Gene Name
F-box and WD repeat domain containing 4
Gene Aliases
DAC, FBW4, FBWD4, SHFM3, SHSF3
UniGene
Hs.500822
Gene Chromosome Location
Chr.10: 101610664-101695295 on Build GRCh38
Transcripts
NM_001323541.1 NM_022039.3 NR_136613.1 XM_005270053.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6413823 c.1237T>A p.*413R Chr.10: 101611293-101611293 on Build GRCh38
5671543 c.1144C>A p.L382I Chr.10: 101611386-101611386 on Build GRCh38
200170 c.1137G>A p.S379S Chr.10: 101611393-101611393 on Build GRCh38
1206876 c.1216G>A p.V406I Chr.10: 101611314-101611314 on Build GRCh38
4011143 c.1135T>A p.S379T Chr.10: 101611395-101611395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922376579 Chr.10: 101611340-101611340 on Build GRCh38
rs369647099 Chr.10: 101611440-101611440 on Build GRCh38
rs765268934 Chr.10: 101611397-101611397 on Build GRCh38
rs781525062 Chr.10: 101611441-101611441 on Build GRCh38
rs879389980 Chr.10: 101611412-101611412 on Build GRCh38
rs771226213 Chr.10: 101611367-101611367 on Build GRCh38
rs753881830 Chr.10: 101611394-101611394 on Build GRCh38
rs574754923 Chr.10: 101611396-101611396 on Build GRCh38
rs371569418 Chr.10: 101611420-101611420 on Build GRCh38
rs778569309 Chr.10: 101611362-101611362 on Build GRCh38
rs142121812 Chr.10: 101611320-101611320 on Build GRCh38
rs780222598 Chr.10: 101611275-101611276 on Build GRCh38
rs748394831 Chr.10: 101611248-101611248 on Build GRCh38
rs376329921 Chr.10: 101611289-101611289 on Build GRCh38
rs200476624 Chr.10: 101611338-101611338 on Build GRCh38
rs749803824 Chr.10: 101611344-101611345 on Build GRCh38
rs753773093 Chr.10: 101611322-101611322 on Build GRCh38
rs770305226 Chr.10: 101611444-101611444 on Build GRCh38
rs777074009 Chr.10: 101611288-101611288 on Build GRCh38
rs761207534 Chr.10: 101611279-101611279 on Build GRCh38
rs934133240 Chr.10: 101611344-101611344 on Build GRCh38
rs970006258 Chr.10: 101611293-101611293 on Build GRCh38
rs773599578 Chr.10: 101611275-101611275 on Build GRCh38
rs148012023 Chr.10: 101611361-101611361 on Build GRCh38
rs751239309 Chr.10: 101611294-101611294 on Build GRCh38
rs577315355 Chr.10: 101611371-101611371 on Build GRCh38
rs747166203 Chr.10: 101611429-101611429 on Build GRCh38
rs772376608 Chr.10: 101611272-101611272 on Build GRCh38
rs556578828 Chr.10: 101611393-101611393 on Build GRCh38
rs750201843 Chr.10: 101611314-101611314 on Build GRCh38
rs371354165 Chr.10: 101611309-101611309 on Build GRCh38
rs143749431 Chr.10: 101611348-101611348 on Build GRCh38
rs755116504 Chr.10: 101611335-101611335 on Build GRCh38
rs939713967 Chr.10: 101611324-101611324 on Build GRCh38
rs959842190 Chr.10: 101611229-101611229 on Build GRCh38
rs747395437 Chr.10: 101611363-101611363 on Build GRCh38
rs763592228 Chr.10: 101611290-101611290 on Build GRCh38
rs758359860 Chr.10: 101611403-101611403 on Build GRCh38
rs779019249 Chr.10: 101611246-101611246 on Build GRCh38
rs761514890 Chr.10: 101611303-101611303 on Build GRCh38
rs35209506 Chr.10: 101611262-101611263 on Build GRCh38
rs73349895 Chr.10: 101611284-101611284 on Build GRCh38
rs902500150 Chr.10: 101611404-101611404 on Build GRCh38
rs761413112 Chr.10: 101611387-101611387 on Build GRCh38
rs755936627 Chr.10: 101611318-101611318 on Build GRCh38
rs992678196 Chr.10: 101611258-101611258 on Build GRCh38
rs542215742 Chr.10: 101611402-101611402 on Build GRCh38
rs537956623 Chr.10: 101611388-101611388 on Build GRCh38
rs867825477 Chr.10: 101611235-101611235 on Build GRCh38
rs773889798 Chr.10: 101611383-101611383 on Build GRCh38
rs760562724 Chr.10: 101611389-101611389 on Build GRCh38
rs368185192 Chr.10: 101611375-101611375 on Build GRCh38

View Full Product Details