Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64088602-64088845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM96A
Entrez Gene ID
84191
Gene Name
family with sequence similarity 96 member A
Gene Aliases
-
UniGene
Hs.439548
Gene Chromosome Location
Chr.15: 64072559-64094008 on Build GRCh38
Transcripts
NM_001014812.2 NM_001289108.1 NM_032231.5 NR_110310.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
138971 c.151G>C p.E51Q Chr.15: 64088825-64088825 on Build GRCh38
470948 c.183G>T p.V61V Chr.15: 64088793-64088793 on Build GRCh38
4683309 c.188C>T p.S63L Chr.15: 64088788-64088788 on Build GRCh38
5640510 c.210G>A p.Q70Q Chr.15: 64088766-64088766 on Build GRCh38
1301315 c.220G>A p.E74K Chr.15: 64088756-64088756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548248324 Chr.15: 64088697-64088697 on Build GRCh38
rs370841346 Chr.15: 64088666-64088666 on Build GRCh38
rs1057395143 Chr.15: 64088670-64088670 on Build GRCh38
rs138828399 Chr.15: 64088742-64088742 on Build GRCh38
rs749545370 Chr.15: 64088790-64088790 on Build GRCh38
rs764870264 Chr.15: 64088667-64088667 on Build GRCh38
rs764039761 Chr.15: 64088748-64088748 on Build GRCh38
rs757810258 Chr.15: 64088794-64088794 on Build GRCh38
rs779631916 Chr.15: 64088702-64088702 on Build GRCh38
rs986233803 Chr.15: 64088718-64088718 on Build GRCh38
rs745890561 Chr.15: 64088738-64088738 on Build GRCh38
rs1027228149 Chr.15: 64088758-64088758 on Build GRCh38
rs757805784 Chr.15: 64088777-64088777 on Build GRCh38
rs756509353 Chr.15: 64088723-64088723 on Build GRCh38
rs749860603 Chr.15: 64088725-64088725 on Build GRCh38
rs369101440 Chr.15: 64088735-64088735 on Build GRCh38
rs1027992794 Chr.15: 64088676-64088676 on Build GRCh38
rs760885827 Chr.15: 64088699-64088699 on Build GRCh38
rs760959396 Chr.15: 64088745-64088745 on Build GRCh38
rs749836777 Chr.15: 64088671-64088671 on Build GRCh38
rs776564285 Chr.15: 64088751-64088751 on Build GRCh38
rs953817117 Chr.15: 64088710-64088710 on Build GRCh38
rs761815245 Chr.15: 64088762-64088762 on Build GRCh38
rs748542366 Chr.15: 64088720-64088720 on Build GRCh38
rs772329697 Chr.15: 64088816-64088816 on Build GRCh38
rs778086456 Chr.15: 64088724-64088724 on Build GRCh38
rs765026006 Chr.15: 64088764-64088764 on Build GRCh38
rs754609426 Chr.15: 64088787-64088787 on Build GRCh38
rs147180860 Chr.15: 64088736-64088736 on Build GRCh38
rs779252477 Chr.15: 64088809-64088809 on Build GRCh38
rs142829471 Chr.15: 64088813-64088813 on Build GRCh38
rs778174210 Chr.15: 64088788-64088788 on Build GRCh38
rs749970749 Chr.15: 64088771-64088771 on Build GRCh38
rs765841610 Chr.15: 64088782-64088782 on Build GRCh38
rs962686932 Chr.15: 64088643-64088643 on Build GRCh38
rs751259047 Chr.15: 64088783-64088783 on Build GRCh38
rs758082475 Chr.15: 64088695-64088695 on Build GRCh38

View Full Product Details