Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134277285-134277524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLB1L3
Entrez Gene ID
112937
Gene Name
galactosidase beta 1 like 3
Gene Aliases
-
UniGene
Hs.721382
Gene Chromosome Location
Chr.11: 134275447-134321364 on Build GRCh38
Transcripts
NM_001080407.2 XM_011542566.2 XM_011542567.2 XM_011542568.2 XM_011542569.2 XM_011542570.2 XM_011542571.2 XM_017017155.1 XM_017017156.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6997125 c.138G>A p.P46P Chr.11: 134277440-134277440 on Build GRCh38
4524208 c.125G>A p.G42E Chr.11: 134277427-134277427 on Build GRCh38
4145552 c.95G>T p.R32L Chr.11: 134277397-134277397 on Build GRCh38
4145551 c.95G>T p.R32L Chr.11: 134277397-134277397 on Build GRCh38
7180823 c.132G>A p.A44A Chr.11: 134277434-134277434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777398392 Chr.11: 134277379-134277379 on Build GRCh38
rs377753637 Chr.11: 134277307-134277307 on Build GRCh38
rs762652301 Chr.11: 134277414-134277415 on Build GRCh38
rs200906355 Chr.11: 134277402-134277402 on Build GRCh38
rs769343043 Chr.11: 134277377-134277377 on Build GRCh38
rs765889873 Chr.11: 134277446-134277449 on Build GRCh38
rs761332446 Chr.11: 134277370-134277371 on Build GRCh38
rs777245399 Chr.11: 134277405-134277405 on Build GRCh38
rs879192305 Chr.11: 134277306-134277306 on Build GRCh38
rs773634038 Chr.11: 134277496-134277496 on Build GRCh38
rs769226039 Chr.11: 134277304-134277304 on Build GRCh38
rs1031103809 Chr.11: 134277353-134277353 on Build GRCh38
rs199729217 Chr.11: 134277420-134277420 on Build GRCh38
rs35657928 Chr.11: 134277389-134277389 on Build GRCh38
rs765897137 Chr.11: 134277331-134277331 on Build GRCh38
rs746129810 Chr.11: 134277383-134277383 on Build GRCh38
rs759319866 Chr.11: 134277318-134277318 on Build GRCh38
rs747016662 Chr.11: 134277397-134277397 on Build GRCh38
rs751014594 Chr.11: 134277354-134277354 on Build GRCh38
rs67473835 Chr.11: 134277477-134277477 on Build GRCh38
rs752562920 Chr.11: 134277371-134277371 on Build GRCh38
rs750134337 Chr.11: 134277328-134277328 on Build GRCh38
rs987685428 Chr.11: 134277433-134277433 on Build GRCh38
rs1029190654 Chr.11: 134277417-134277417 on Build GRCh38
rs556952102 Chr.11: 134277444-134277444 on Build GRCh38
rs748715795 Chr.11: 134277490-134277490 on Build GRCh38
rs765952222 Chr.11: 134277422-134277422 on Build GRCh38
rs773986215 Chr.11: 134277424-134277424 on Build GRCh38
rs762299190 Chr.11: 134277311-134277311 on Build GRCh38
rs781255682 Chr.11: 134277365-134277365 on Build GRCh38
rs755984875 Chr.11: 134277378-134277378 on Build GRCh38
rs754916305 Chr.11: 134277356-134277356 on Build GRCh38
rs755964881 Chr.11: 134277443-134277443 on Build GRCh38
rs200184626 Chr.11: 134277411-134277411 on Build GRCh38
rs201676488 Chr.11: 134277485-134277485 on Build GRCh38
rs758696157 Chr.11: 134277467-134277467 on Build GRCh38
rs766911388 Chr.11: 134277435-134277435 on Build GRCh38
rs762611229 Chr.11: 134277415-134277415 on Build GRCh38
rs374355226 Chr.11: 134277410-134277410 on Build GRCh38
rs764964077 Chr.11: 134277394-134277394 on Build GRCh38
rs575307611 Chr.11: 134277472-134277472 on Build GRCh38
rs978636511 Chr.11: 134277476-134277476 on Build GRCh38
rs780291464 Chr.11: 134277390-134277390 on Build GRCh38
rs772676676 Chr.11: 134277411-134277412 on Build GRCh38
rs753628808 Chr.11: 134277449-134277449 on Build GRCh38
rs748573471 Chr.11: 134277408-134277408 on Build GRCh38
rs199737816 Chr.11: 134277439-134277439 on Build GRCh38
rs758640988 Chr.11: 134277386-134277386 on Build GRCh38
rs759102239 Chr.11: 134277434-134277434 on Build GRCh38

View Full Product Details