Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10874069-10874328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCM2
Entrez Gene ID
9247
Gene Name
glial cells missing homolog 2
Gene Aliases
GCMB, hGCMb
UniGene
Hs.227098
Gene Chromosome Location
Chr.6: 10873223-10881865 on Build GRCh38
Transcripts
NM_004752.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1696925 c.1406C>T p.S469F Chr.6: 10874110-10874110 on Build GRCh38
6842719 c.1348A>T p.I450F Chr.6: 10874168-10874168 on Build GRCh38
5634593 c.1428G>A p.E476E Chr.6: 10874088-10874088 on Build GRCh38
1696926 c.1355G>A p.G452E Chr.6: 10874161-10874161 on Build GRCh38
5657395 c.1189C>A p.P397T Chr.6: 10874327-10874327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145034037 Chr.6: 10874299-10874299 on Build GRCh38
rs767662821 Chr.6: 10874252-10874252 on Build GRCh38
rs139538731 Chr.6: 10874121-10874121 on Build GRCh38
rs182776740 Chr.6: 10874257-10874257 on Build GRCh38
rs749101028 Chr.6: 10874237-10874237 on Build GRCh38
rs200926987 Chr.6: 10874218-10874218 on Build GRCh38
rs779346724 Chr.6: 10874103-10874103 on Build GRCh38
rs142084903 Chr.6: 10874216-10874216 on Build GRCh38
rs745544582 Chr.6: 10874120-10874120 on Build GRCh38
rs527620122 Chr.6: 10874217-10874217 on Build GRCh38
rs147308895 Chr.6: 10874106-10874106 on Build GRCh38
rs140246165 Chr.6: 10874180-10874180 on Build GRCh38
rs956579123 Chr.6: 10874199-10874200 on Build GRCh38
rs958487240 Chr.6: 10874243-10874243 on Build GRCh38
rs756711799 Chr.6: 10874210-10874210 on Build GRCh38
rs761953054 Chr.6: 10874155-10874155 on Build GRCh38
rs765546248 Chr.6: 10874271-10874271 on Build GRCh38
rs145103332 Chr.6: 10874152-10874152 on Build GRCh38
rs748030466 Chr.6: 10874300-10874300 on Build GRCh38
rs769221323 Chr.6: 10874092-10874092 on Build GRCh38
rs537546612 Chr.6: 10874118-10874118 on Build GRCh38
rs750622385 Chr.6: 10874173-10874173 on Build GRCh38
rs755365377 Chr.6: 10874198-10874198 on Build GRCh38
rs146478181 Chr.6: 10874281-10874281 on Build GRCh38
rs768448554 Chr.6: 10874294-10874294 on Build GRCh38
rs775355911 Chr.6: 10874126-10874126 on Build GRCh38
rs387907433 Chr.6: 10874239-10874239 on Build GRCh38
rs1037580555 Chr.6: 10874157-10874157 on Build GRCh38
rs769274226 Chr.6: 10874176-10874176 on Build GRCh38
rs149925208 Chr.6: 10874280-10874280 on Build GRCh38
rs770275682 Chr.6: 10874214-10874214 on Build GRCh38
rs754219196 Chr.6: 10874190-10874190 on Build GRCh38
rs141025277 Chr.6: 10874286-10874286 on Build GRCh38
rs753165002 Chr.6: 10874272-10874272 on Build GRCh38
rs774323324 Chr.6: 10874238-10874238 on Build GRCh38
rs140411555 Chr.6: 10874266-10874266 on Build GRCh38
rs989589336 Chr.6: 10874295-10874295 on Build GRCh38
rs772123246 Chr.6: 10874301-10874301 on Build GRCh38
rs577206471 Chr.6: 10874201-10874201 on Build GRCh38
rs1017017909 Chr.6: 10874255-10874255 on Build GRCh38
rs774218255 Chr.6: 10874153-10874153 on Build GRCh38
rs371458608 Chr.6: 10874287-10874287 on Build GRCh38
rs147168542 Chr.6: 10874174-10874174 on Build GRCh38
rs767754568 Chr.6: 10874164-10874164 on Build GRCh38
rs771107707 Chr.6: 10874269-10874269 on Build GRCh38
rs1002762397 Chr.6: 10874224-10874224 on Build GRCh38
rs760897802 Chr.6: 10874264-10874264 on Build GRCh38
rs1054175807 Chr.6: 10874172-10874172 on Build GRCh38
rs555770771 Chr.6: 10874140-10874140 on Build GRCh38
rs567065650 Chr.6: 10874115-10874115 on Build GRCh38
rs748941572 Chr.6: 10874292-10874292 on Build GRCh38
rs151218849 Chr.6: 10874229-10874229 on Build GRCh38

View Full Product Details