Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33915599-33915870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5O
Entrez Gene ID
539
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit
Gene Aliases
ATPO, HMC08D05, OSCP
UniGene
Hs.409140
Gene Chromosome Location
Chr.21: 33903453-33915854 on Build GRCh38
Transcripts
NM_001697.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3972762 c.21C>T p.S7S Chr.21: 33915743-33915743 on Build GRCh38
6161374 c.14C>T p.A5V Chr.21: 33915750-33915750 on Build GRCh38
4388936 c.14C>G p.A5G Chr.21: 33915750-33915750 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750826555 Chr.21: 33915802-33915802 on Build GRCh38
rs112857598 Chr.21: 33915813-33915813 on Build GRCh38
rs201852944 Chr.21: 33915745-33915745 on Build GRCh38
rs896967780 Chr.21: 33915743-33915743 on Build GRCh38
rs772092840 Chr.21: 33915811-33915811 on Build GRCh38
rs779738813 Chr.21: 33915799-33915799 on Build GRCh38
rs771384495 Chr.21: 33915804-33915804 on Build GRCh38
rs771863868 Chr.21: 33915633-33915633 on Build GRCh38
rs770240406 Chr.21: 33915753-33915753 on Build GRCh38
rs868418588 Chr.21: 33915688-33915688 on Build GRCh38
rs756618079 Chr.21: 33915806-33915806 on Build GRCh38
rs922095549 Chr.21: 33915831-33915831 on Build GRCh38
rs144779561 Chr.21: 33915740-33915740 on Build GRCh38
rs1043209649 Chr.21: 33915714-33915714 on Build GRCh38
rs146520965 Chr.21: 33915730-33915730 on Build GRCh38
rs947554103 Chr.21: 33915680-33915680 on Build GRCh38
rs781740423 Chr.21: 33915744-33915744 on Build GRCh38
rs202191698 Chr.21: 33915757-33915757 on Build GRCh38
rs558937660 Chr.21: 33915801-33915801 on Build GRCh38
rs760629150 Chr.21: 33915773-33915773 on Build GRCh38
rs775367269 Chr.21: 33915772-33915772 on Build GRCh38
rs913386536 Chr.21: 33915640-33915640 on Build GRCh38
rs989042790 Chr.21: 33915822-33915822 on Build GRCh38
rs767350855 Chr.21: 33915732-33915732 on Build GRCh38
rs376092149 Chr.21: 33915782-33915782 on Build GRCh38
rs770293793 Chr.21: 33915686-33915686 on Build GRCh38
rs996402266 Chr.21: 33915676-33915676 on Build GRCh38
rs753603487 Chr.21: 33915780-33915780 on Build GRCh38
rs753443463 Chr.21: 33915738-33915738 on Build GRCh38
rs763296469 Chr.21: 33915703-33915703 on Build GRCh38
rs750101329 Chr.21: 33915795-33915795 on Build GRCh38
rs140977810 Chr.21: 33915733-33915733 on Build GRCh38
rs577087774 Chr.21: 33915809-33915809 on Build GRCh38
rs541443960 Chr.21: 33915645-33915645 on Build GRCh38
rs959993523 Chr.21: 33915810-33915810 on Build GRCh38
rs746132315 Chr.21: 33915807-33915807 on Build GRCh38
rs947601107 Chr.21: 33915654-33915654 on Build GRCh38
rs538892695 Chr.21: 33915627-33915627 on Build GRCh38
rs928931479 Chr.21: 33915630-33915630 on Build GRCh38
rs762923806 Chr.21: 33915711-33915711 on Build GRCh38
rs9979313 Chr.21: 33915661-33915661 on Build GRCh38
rs757777401 Chr.21: 33915796-33915796 on Build GRCh38
rs756841691 Chr.21: 33915739-33915739 on Build GRCh38
rs938379054 Chr.21: 33915628-33915628 on Build GRCh38
rs775844351 Chr.21: 33915670-33915670 on Build GRCh38
rs764281169 Chr.21: 33915712-33915712 on Build GRCh38
rs932201408 Chr.21: 33915836-33915836 on Build GRCh38
rs369688647 Chr.21: 33915755-33915755 on Build GRCh38
rs759477820 Chr.21: 33915770-33915770 on Build GRCh38
rs774700870 Chr.21: 33915764-33915764 on Build GRCh38
rs780451873 Chr.21: 33915683-33915683 on Build GRCh38
rs774394655 Chr.21: 33915800-33915801 on Build GRCh38
rs778462106 Chr.21: 33915803-33915803 on Build GRCh38
rs762435556 Chr.21: 33915767-33915767 on Build GRCh38
rs372840826 Chr.21: 33915775-33915775 on Build GRCh38
rs377315164 Chr.21: 33915734-33915734 on Build GRCh38
rs978929539 Chr.21: 33915829-33915829 on Build GRCh38
rs763907878 Chr.21: 33915737-33915737 on Build GRCh38
rs770992501 Chr.21: 33915713-33915713 on Build GRCh38
rs773498022 Chr.21: 33915694-33915694 on Build GRCh38

View Full Product Details