Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19645514-19645749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A1
Entrez Gene ID
57130
Gene Name
ATPase 13A1
Gene Aliases
ATP13A, CGI-152
UniGene
Hs.501794
Gene Chromosome Location
Chr.19: 19645198-19663694 on Build GRCh38
Transcripts
NM_020410.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4936747 c.3402G>A p.L1134L Chr.19: 19645749-19645749 on Build GRCh38
6432515 c.3410G>T p.S1137I Chr.19: 19645741-19645741 on Build GRCh38
4936748 c.3048G>A p.L1016L Chr.19: 19645749-19645749 on Build GRCh38
1196321 c.3470A>G p.N1157S Chr.19: 19645681-19645681 on Build GRCh38
5421476 c.3463G>T p.D1155Y Chr.19: 19645688-19645688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779478989 Chr.19: 19645652-19645652 on Build GRCh38
rs780873055 Chr.19: 19645581-19645581 on Build GRCh38
rs890576648 Chr.19: 19645564-19645564 on Build GRCh38
rs755592112 Chr.19: 19645727-19645727 on Build GRCh38
rs764001956 Chr.19: 19645693-19645693 on Build GRCh38
rs748523704 Chr.19: 19645656-19645656 on Build GRCh38
rs749826991 Chr.19: 19645634-19645634 on Build GRCh38
rs200525190 Chr.19: 19645537-19645537 on Build GRCh38
rs994692568 Chr.19: 19645658-19645658 on Build GRCh38
rs749189208 Chr.19: 19645706-19645706 on Build GRCh38
rs368332372 Chr.19: 19645565-19645565 on Build GRCh38
rs1050527971 Chr.19: 19645562-19645562 on Build GRCh38
rs372187284 Chr.19: 19645716-19645716 on Build GRCh38
rs771015526 Chr.19: 19645686-19645686 on Build GRCh38
rs762949014 Chr.19: 19645689-19645689 on Build GRCh38
rs756913302 Chr.19: 19645579-19645579 on Build GRCh38
rs774058874 Chr.19: 19645701-19645701 on Build GRCh38
rs779659570 Chr.19: 19645620-19645620 on Build GRCh38
rs1027556673 Chr.19: 19645691-19645691 on Build GRCh38
rs542153953 Chr.19: 19645541-19645541 on Build GRCh38
rs374676055 Chr.19: 19645704-19645704 on Build GRCh38
rs374619003 Chr.19: 19645542-19645542 on Build GRCh38
rs563912098 Chr.19: 19645688-19645688 on Build GRCh38
rs370911404 Chr.19: 19645538-19645538 on Build GRCh38
rs747153287 Chr.19: 19645681-19645681 on Build GRCh38
rs772224519 Chr.19: 19645665-19645665 on Build GRCh38
rs367952388 Chr.19: 19645714-19645714 on Build GRCh38
rs755468322 Chr.19: 19645639-19645639 on Build GRCh38
rs748387676 Chr.19: 19645692-19645692 on Build GRCh38
rs767885866 Chr.19: 19645537-19645537 on Build GRCh38
rs773515667 Chr.19: 19645679-19645679 on Build GRCh38
rs201864782 Chr.19: 19645614-19645614 on Build GRCh38
rs749938947 Chr.19: 19645611-19645611 on Build GRCh38
rs750406558 Chr.19: 19645627-19645644 on Build GRCh38
rs755663907 Chr.19: 19645710-19645710 on Build GRCh38
rs191656095 Chr.19: 19645588-19645588 on Build GRCh38
rs550479637 Chr.19: 19645622-19645622 on Build GRCh38

View Full Product Details