Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200815260-200815485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BZW1
Entrez Gene ID
9689
Gene Name
basic leucine zipper and W2 domains 1
Gene Aliases
BZAP45, Nbla10236
UniGene
Hs.355983
Gene Chromosome Location
Chr.2: 200811546-200823851 on Build GRCh38
Transcripts
NM_001207067.1 NM_001207069.1 NM_001321688.1 NM_001321690.1 NM_001321691.1 NM_001321693.1 NM_001321694.1 NM_014670.3 XM_017005393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6613758 c.161-2A>G p.? Chr.2: 200815339-200815339 on Build GRCh38
3425933 c.290G>A p.R97Q Chr.2: 200815470-200815470 on Build GRCh38
3938866 c.203T>C p.I68T Chr.2: 200815383-200815383 on Build GRCh38
5142580 c.116G>A p.G39D Chr.2: 200815392-200815392 on Build GRCh38
7296646 c.190C>T p.R64C Chr.2: 200815466-200815466 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763170670 Chr.2: 200815431-200815431 on Build GRCh38
rs752893493 Chr.2: 200815328-200815328 on Build GRCh38
rs11550649 Chr.2: 200815459-200815459 on Build GRCh38
rs546040551 Chr.2: 200815324-200815324 on Build GRCh38
rs758960828 Chr.2: 200815329-200815329 on Build GRCh38
rs748078736 Chr.2: 200815301-200815301 on Build GRCh38
rs754415074 Chr.2: 200815380-200815380 on Build GRCh38
rs777611664 Chr.2: 200815398-200815398 on Build GRCh38
rs759835220 Chr.2: 200815414-200815414 on Build GRCh38
rs551505298 Chr.2: 200815384-200815384 on Build GRCh38
rs908571197 Chr.2: 200815331-200815331 on Build GRCh38
rs974595283 Chr.2: 200815460-200815460 on Build GRCh38
rs779710956 Chr.2: 200815369-200815369 on Build GRCh38
rs762977368 Chr.2: 200815312-200815313 on Build GRCh38
rs751926132 Chr.2: 200815340-200815340 on Build GRCh38
rs368021486 Chr.2: 200815376-200815376 on Build GRCh38
rs774730544 Chr.2: 200815407-200815408 on Build GRCh38
rs772060333 Chr.2: 200815302-200815302 on Build GRCh38
rs765414539 Chr.2: 200815327-200815327 on Build GRCh38
rs151292360 Chr.2: 200815323-200815323 on Build GRCh38
rs372199927 Chr.2: 200815409-200815409 on Build GRCh38
rs759638323 Chr.2: 200815326-200815326 on Build GRCh38
rs746778000 Chr.2: 200815403-200815403 on Build GRCh38
rs770199814 Chr.2: 200815415-200815415 on Build GRCh38
rs757597354 Chr.2: 200815359-200815359 on Build GRCh38
rs771970525 Chr.2: 200815397-200815397 on Build GRCh38
rs766591961 Chr.2: 200815316-200815316 on Build GRCh38
rs770652522 Chr.2: 200815405-200815405 on Build GRCh38
rs773042803 Chr.2: 200815305-200815305 on Build GRCh38
rs764557922 Chr.2: 200815333-200815333 on Build GRCh38
rs764626498 Chr.2: 200815447-200815447 on Build GRCh38
rs569745449 Chr.2: 200815426-200815426 on Build GRCh38
rs747710315 Chr.2: 200815388-200815388 on Build GRCh38
rs771118807 Chr.2: 200815318-200815338 on Build GRCh38

View Full Product Details