Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108229209-108229434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATM
Entrez Gene ID
472
Gene Name
ATM serine/threonine kinase
Gene Aliases
AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1
UniGene
Hs.367437
Gene Chromosome Location
Chr.11: 108222484-108369102 on Build GRCh38
Transcripts
NM_000051.3 XM_005271561.4 XM_005271562.4 XM_006718843.3 XM_011542840.2 XM_011542842.2 XM_011542843.2 XM_011542844.2 XM_017017789.1 XM_017017790.1 XM_017017791.1 XM_017017792.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5506968 c.307T>G p.Y103D Chr.11: 108229299-108229299 on Build GRCh38
3979082 c.231A>T p.I77I Chr.11: 108229223-108229223 on Build GRCh38
6978980 c.324_331delAAACAGAA p.N109fs*3 Chr.11: 108229316-108229323 on Build GRCh38
3808274 c.315C>T p.I105I Chr.11: 108229307-108229307 on Build GRCh38
922697 c.299T>G p.L100W Chr.11: 108229291-108229291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753349239 Chr.11: 108229379-108229381 on Build GRCh38
rs761936549 Chr.11: 108229291-108229291 on Build GRCh38
rs879254161 Chr.11: 108229291-108229299 on Build GRCh38
rs137882485 Chr.11: 108229287-108229287 on Build GRCh38
rs201887139 Chr.11: 108229365-108229365 on Build GRCh38
rs587781937 Chr.11: 108229243-108229243 on Build GRCh38
rs552244152 Chr.11: 108229360-108229360 on Build GRCh38
rs777499935 Chr.11: 108229301-108229301 on Build GRCh38
rs756969590 Chr.11: 108229266-108229266 on Build GRCh38
rs759952043 Chr.11: 108229374-108229374 on Build GRCh38
rs765562938 Chr.11: 108229377-108229378 on Build GRCh38
rs114106540 Chr.11: 108229378-108229378 on Build GRCh38
rs786201137 Chr.11: 108229268-108229268 on Build GRCh38
rs189555352 Chr.11: 108229372-108229372 on Build GRCh38
rs530540420 Chr.11: 108229351-108229351 on Build GRCh38
rs142358238 Chr.11: 108229312-108229312 on Build GRCh38
rs375605135 Chr.11: 108229258-108229258 on Build GRCh38
rs755557468 Chr.11: 108229333-108229333 on Build GRCh38
rs56324032 Chr.11: 108229371-108229371 on Build GRCh38
rs777434093 Chr.11: 108229250-108229250 on Build GRCh38
rs878853503 Chr.11: 108229331-108229331 on Build GRCh38
rs200151849 Chr.11: 108229267-108229267 on Build GRCh38
rs397713128 Chr.11: 108229392-108229393 on Build GRCh38
rs752156866 Chr.11: 108229379-108229379 on Build GRCh38
rs766951228 Chr.11: 108229315-108229315 on Build GRCh38
rs746762110 Chr.11: 108229283-108229283 on Build GRCh38
rs868748140 Chr.11: 108229260-108229260 on Build GRCh38
rs764823818 Chr.11: 108229379-108229379 on Build GRCh38
rs750761029 Chr.11: 108229378-108229378 on Build GRCh38
rs776276698 Chr.11: 108229368-108229368 on Build GRCh38
rs11390378 Chr.11: 108229378-108229379 on Build GRCh38
rs878853501 Chr.11: 108229309-108229309 on Build GRCh38
rs377422031 Chr.11: 108229350-108229350 on Build GRCh38
rs78327881 Chr.11: 108229393-108229393 on Build GRCh38
rs750059218 Chr.11: 108229381-108229381 on Build GRCh38
rs368196317 Chr.11: 108229271-108229271 on Build GRCh38
rs878853499 Chr.11: 108229292-108229292 on Build GRCh38
rs879225002 Chr.11: 108229363-108229363 on Build GRCh38
rs752135143 Chr.11: 108229328-108229328 on Build GRCh38
rs541933218 Chr.11: 108229348-108229348 on Build GRCh38
rs372974993 Chr.11: 108229373-108229373 on Build GRCh38
rs864622508 Chr.11: 108229311-108229311 on Build GRCh38
rs587781545 Chr.11: 108229275-108229275 on Build GRCh38
rs878998551 Chr.11: 108229377-108229377 on Build GRCh38
rs864622758 Chr.11: 108229272-108229272 on Build GRCh38
rs768318076 Chr.11: 108229284-108229284 on Build GRCh38
rs767739059 Chr.11: 108229334-108229334 on Build GRCh38
rs758483894 Chr.11: 108229293-108229293 on Build GRCh38
rs878853497 Chr.11: 108229281-108229281 on Build GRCh38
rs778546616 Chr.11: 108229345-108229345 on Build GRCh38
rs786203011 Chr.11: 108229282-108229282 on Build GRCh38
rs730881370 Chr.11: 108229314-108229314 on Build GRCh38
rs756843866 Chr.11: 108229340-108229340 on Build GRCh38
rs755326770 Chr.11: 108229249-108229249 on Build GRCh38
rs756535279 Chr.11: 108229376-108229376 on Build GRCh38
rs753168614 Chr.11: 108229338-108229338 on Build GRCh38
rs139351982 Chr.11: 108229362-108229362 on Build GRCh38

View Full Product Details