Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 143436772-143437031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP15
Entrez Gene ID
55843
Gene Name
Rho GTPase activating protein 15
Gene Aliases
BM046
UniGene
Hs.171011
Gene Chromosome Location
Chr.2: 143070973-143768352 on Build GRCh38
Transcripts
NM_018460.3 XM_011511479.2 XM_011511481.2 XM_011511482.2 XM_011511483.1 XM_011511484.1 XM_017004499.1 XM_017004500.1 XM_017004501.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1305677 c.607G>C p.E203Q Chr.2: 143436946-143436946 on Build GRCh38
3757656 c.657C>T p.Y219Y Chr.2: 143436996-143436996 on Build GRCh38
309232 c.603C>T p.N201N Chr.2: 143436942-143436942 on Build GRCh38
6038354 c.691A>G p.R231G Chr.2: 143437030-143437030 on Build GRCh38
1305678 c.640G>A p.E214K Chr.2: 143436979-143436979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774313685 Chr.2: 143436942-143436942 on Build GRCh38
rs759348173 Chr.2: 143436943-143436943 on Build GRCh38
rs755585857 Chr.2: 143436867-143436867 on Build GRCh38
rs747120568 Chr.2: 143436915-143436915 on Build GRCh38
rs768765146 Chr.2: 143436921-143436921 on Build GRCh38
rs369879273 Chr.2: 143436861-143436861 on Build GRCh38
rs748298682 Chr.2: 143436866-143436869 on Build GRCh38
rs17230746 Chr.2: 143436996-143436996 on Build GRCh38
rs1037706819 Chr.2: 143436876-143436876 on Build GRCh38
rs763722517 Chr.2: 143436877-143436877 on Build GRCh38
rs375998378 Chr.2: 143436976-143436976 on Build GRCh38
rs775494512 Chr.2: 143436972-143436972 on Build GRCh38
rs961193230 Chr.2: 143436847-143436848 on Build GRCh38
rs564252160 Chr.2: 143436842-143436842 on Build GRCh38
rs886347587 Chr.2: 143436840-143436840 on Build GRCh38
rs923080648 Chr.2: 143436814-143436814 on Build GRCh38
rs770972369 Chr.2: 143436798-143436798 on Build GRCh38
rs748353571 Chr.2: 143436932-143436932 on Build GRCh38
rs763629028 Chr.2: 143436977-143436977 on Build GRCh38
rs758916393 Chr.2: 143436896-143436896 on Build GRCh38
rs753247677 Chr.2: 143436989-143436989 on Build GRCh38
rs1047577999 Chr.2: 143436831-143436831 on Build GRCh38
rs199632589 Chr.2: 143436936-143436936 on Build GRCh38
rs1038760817 Chr.2: 143436808-143436808 on Build GRCh38
rs767566594 Chr.2: 143436951-143436951 on Build GRCh38
rs776771477 Chr.2: 143436924-143436924 on Build GRCh38
rs779278702 Chr.2: 143436892-143436892 on Build GRCh38
rs1045312664 Chr.2: 143436955-143436955 on Build GRCh38
rs6755050 Chr.2: 143436878-143436878 on Build GRCh38
rs531723191 Chr.2: 143436848-143436848 on Build GRCh38
rs780541707 Chr.2: 143436913-143436913 on Build GRCh38
rs763352524 Chr.2: 143436813-143436813 on Build GRCh38
rs757657914 Chr.2: 143436884-143436884 on Build GRCh38
rs545848542 Chr.2: 143436838-143436838 on Build GRCh38
rs933173727 Chr.2: 143436827-143436827 on Build GRCh38

View Full Product Details