Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 170234752-170235023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf58
Entrez Gene ID
133874
Gene Name
chromosome 5 open reading frame 58
Gene Aliases
-
UniGene
Hs.519749
Gene Chromosome Location
Chr.5: 170231730-170252569 on Build GRCh38
Transcripts
NM_001102609.2 NM_001305393.1 NM_001305394.1 NM_001305395.1 NM_001305396.1 NM_001305397.1 NR_131091.1 NR_131092.1 XM_011534432.2 XM_017009029.1 XM_017009030.1 XM_017009031.1 XM_017009032.1 XM_017009033.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7295565 c.76C>T p.R26C Chr.5: 170234989-170234989 on Build GRCh38
3661710 c.93G>A p.K31K Chr.5: 170235006-170235006 on Build GRCh38
5795269 c.77G>A p.R26H Chr.5: 170234990-170234990 on Build GRCh38
6365003 c.92A>G p.K31R Chr.5: 170235005-170235005 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766098383 Chr.5: 170234971-170234985 on Build GRCh38
rs1013744636 Chr.5: 170234878-170234878 on Build GRCh38
rs747777610 Chr.5: 170234961-170234961 on Build GRCh38
rs768143872 Chr.5: 170234950-170234950 on Build GRCh38
rs367926237 Chr.5: 170234929-170234929 on Build GRCh38
rs181994244 Chr.5: 170234916-170234916 on Build GRCh38
rs753014198 Chr.5: 170234819-170234819 on Build GRCh38
rs957999 Chr.5: 170234945-170234945 on Build GRCh38
rs879386174 Chr.5: 170234820-170234820 on Build GRCh38
rs776294546 Chr.5: 170234959-170234959 on Build GRCh38
rs551597672 Chr.5: 170234829-170234829 on Build GRCh38
rs1031132248 Chr.5: 170234797-170234797 on Build GRCh38
rs746413537 Chr.5: 170234928-170234928 on Build GRCh38
rs144652063 Chr.5: 170234794-170234794 on Build GRCh38
rs957998 Chr.5: 170234990-170234990 on Build GRCh38
rs866295314 Chr.5: 170234848-170234848 on Build GRCh38
rs976530010 Chr.5: 170234902-170234902 on Build GRCh38
rs1023747115 Chr.5: 170234882-170234882 on Build GRCh38
rs761217070 Chr.5: 170234885-170234885 on Build GRCh38
rs112927588 Chr.5: 170234843-170234843 on Build GRCh38
rs560558936 Chr.5: 170234947-170234947 on Build GRCh38
rs767944928 Chr.5: 170234981-170234981 on Build GRCh38
rs529764469 Chr.5: 170234809-170234809 on Build GRCh38

View Full Product Details