Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73260824-73261079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF4L
Entrez Gene ID
642843
Gene Name
cleavage and polyadenylation specific factor 4 like
Gene Aliases
-
UniGene
Hs.534707
Gene Chromosome Location
Chr.17: 73242343-73263778 on Build GRCh38
Transcripts
NM_001129885.1 XM_011525115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5791955 c.154+10C>G p.? Chr.17: 73260923-73260923 on Build GRCh38
7362541 c.118G>T p.V40L Chr.17: 73260969-73260969 on Build GRCh38
6317510 c.104-3C>A p.? Chr.17: 73260986-73260986 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368084663 Chr.17: 73261004-73261004 on Build GRCh38
rs568361834 Chr.17: 73260860-73260860 on Build GRCh38
rs754963851 Chr.17: 73260970-73260970 on Build GRCh38
rs996774845 Chr.17: 73260980-73260980 on Build GRCh38
rs926339397 Chr.17: 73260861-73260861 on Build GRCh38
rs779658515 Chr.17: 73260884-73260884 on Build GRCh38
rs899486010 Chr.17: 73261058-73261058 on Build GRCh38
rs557201115 Chr.17: 73260976-73260976 on Build GRCh38
rs746097590 Chr.17: 73261022-73261022 on Build GRCh38
rs892419226 Chr.17: 73260909-73260909 on Build GRCh38
rs199773733 Chr.17: 73260969-73260969 on Build GRCh38
rs982409337 Chr.17: 73260876-73260876 on Build GRCh38
rs939198691 Chr.17: 73260889-73260889 on Build GRCh38
rs761072945 Chr.17: 73261003-73261003 on Build GRCh38
rs1039370542 Chr.17: 73261040-73261040 on Build GRCh38
rs779998731 Chr.17: 73260957-73260957 on Build GRCh38
rs927838550 Chr.17: 73260877-73260877 on Build GRCh38
rs749354815 Chr.17: 73260979-73260979 on Build GRCh38
rs772358651 Chr.17: 73260907-73260907 on Build GRCh38
rs1056669537 Chr.17: 73260894-73260894 on Build GRCh38
rs1009437998 Chr.17: 73260918-73260918 on Build GRCh38
rs766302069 Chr.17: 73260881-73260881 on Build GRCh38

View Full Product Details