Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28857908-28858175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERAL1
Entrez Gene ID
26284
Gene Name
Era like 12S mitochondrial rRNA chaperone 1
Gene Aliases
CEGA, ERA, ERA-W, ERAL1A, H-ERA, HERA-A, HERA-B
UniGene
Hs.3426
Gene Chromosome Location
Chr.17: 28854957-28861067 on Build GRCh38
Transcripts
NM_001317985.1 NM_001317986.1 NM_005702.3 NR_134328.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5781007 c.541C>A p.H181N Chr.17: 28858150-28858150 on Build GRCh38
5414369 c.495A>T p.L165L Chr.17: 28857944-28857944 on Build GRCh38
5901986 c.537-7C>T p.? Chr.17: 28858139-28858139 on Build GRCh38
1742000 c.516C>G p.I172M Chr.17: 28857965-28857965 on Build GRCh38
108591 c.490-1G>A p.? Chr.17: 28857938-28857938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020313033 Chr.17: 28858072-28858072 on Build GRCh38
rs199960158 Chr.17: 28858012-28858012 on Build GRCh38
rs200068552 Chr.17: 28858100-28858100 on Build GRCh38
rs893127595 Chr.17: 28858046-28858046 on Build GRCh38
rs754256288 Chr.17: 28857994-28857994 on Build GRCh38
rs768640741 Chr.17: 28858027-28858027 on Build GRCh38
rs750486213 Chr.17: 28857968-28857968 on Build GRCh38
rs775097680 Chr.17: 28858020-28858020 on Build GRCh38
rs962103123 Chr.17: 28858048-28858048 on Build GRCh38
rs139024598 Chr.17: 28858151-28858151 on Build GRCh38
rs1036267430 Chr.17: 28858135-28858135 on Build GRCh38
rs766323419 Chr.17: 28858120-28858120 on Build GRCh38
rs561462239 Chr.17: 28858068-28858068 on Build GRCh38
rs767700837 Chr.17: 28858096-28858096 on Build GRCh38
rs375134357 Chr.17: 28858016-28858016 on Build GRCh38
rs753085815 Chr.17: 28858109-28858110 on Build GRCh38
rs922422018 Chr.17: 28858079-28858079 on Build GRCh38
rs985356500 Chr.17: 28858150-28858150 on Build GRCh38
rs201563994 Chr.17: 28857936-28857936 on Build GRCh38
rs755535001 Chr.17: 28857998-28857998 on Build GRCh38
rs780574706 Chr.17: 28857992-28857992 on Build GRCh38
rs867504731 Chr.17: 28857965-28857965 on Build GRCh38
rs763747118 Chr.17: 28857941-28857941 on Build GRCh38
rs767792864 Chr.17: 28857955-28857955 on Build GRCh38
rs761955482 Chr.17: 28857946-28857946 on Build GRCh38
rs907923338 Chr.17: 28858037-28858037 on Build GRCh38
rs760985385 Chr.17: 28858114-28858114 on Build GRCh38
rs761651113 Chr.17: 28858036-28858036 on Build GRCh38
rs35872490 Chr.17: 28858015-28858016 on Build GRCh38
rs200777246 Chr.17: 28857999-28857999 on Build GRCh38
rs756928968 Chr.17: 28858021-28858021 on Build GRCh38
rs774128528 Chr.17: 28858031-28858031 on Build GRCh38
rs1051998811 Chr.17: 28857939-28857939 on Build GRCh38
rs753569450 Chr.17: 28857950-28857950 on Build GRCh38
rs756194322 Chr.17: 28857985-28857985 on Build GRCh38
rs146168474 Chr.17: 28857938-28857938 on Build GRCh38
rs1002995516 Chr.17: 28858131-28858131 on Build GRCh38
rs888987799 Chr.17: 28857978-28857978 on Build GRCh38
rs368184082 Chr.17: 28858015-28858015 on Build GRCh38
rs145180494 Chr.17: 28857979-28857979 on Build GRCh38
rs769227873 Chr.17: 28858017-28858017 on Build GRCh38
rs762739233 Chr.17: 28858023-28858023 on Build GRCh38
rs751369228 Chr.17: 28857945-28857945 on Build GRCh38

View Full Product Details