Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 35951642-35951915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP47
Entrez Gene ID
286464
Gene Name
cilia and flagella associated protein 47
Gene Aliases
CHDC2, CXorf22, CXorf30, CXorf59
UniGene
Hs.376425
Gene Chromosome Location
Chr.X: 35919734-36385319 on Build GRCh38
Transcripts
NM_001304548.1 NM_152632.3 XM_017029452.1 XM_017029453.1

SNP Detail

SNP ID Chromosome Location
rs189129830 Chr.X: 35951881-35951881 on Build GRCh38
rs766167088 Chr.X: 35951793-35951793 on Build GRCh38
rs1024512345 Chr.X: 35951837-35951837 on Build GRCh38
rs1010369855 Chr.X: 35951813-35951813 on Build GRCh38
rs375487644 Chr.X: 35951879-35951879 on Build GRCh38
rs200214227 Chr.X: 35951768-35951768 on Build GRCh38
rs956418610 Chr.X: 35951772-35951772 on Build GRCh38
rs890945163 Chr.X: 35951677-35951677 on Build GRCh38
rs776770755 Chr.X: 35951761-35951761 on Build GRCh38
rs1040010564 Chr.X: 35951722-35951722 on Build GRCh38
rs780575676 Chr.X: 35951888-35951888 on Build GRCh38
rs1021045589 Chr.X: 35951740-35951740 on Build GRCh38
rs1006848991 Chr.X: 35951681-35951681 on Build GRCh38
rs369106796 Chr.X: 35951790-35951790 on Build GRCh38
rs773448767 Chr.X: 35951771-35951771 on Build GRCh38
rs752218313 Chr.X: 35951829-35951829 on Build GRCh38
rs755628937 Chr.X: 35951833-35951833 on Build GRCh38
rs143015064 Chr.X: 35951869-35951869 on Build GRCh38
rs767420279 Chr.X: 35951828-35951828 on Build GRCh38
rs980408864 Chr.X: 35951854-35951854 on Build GRCh38
rs759505994 Chr.X: 35951812-35951812 on Build GRCh38
rs1009704481 Chr.X: 35951700-35951700 on Build GRCh38
rs774460927 Chr.X: 35951725-35951725 on Build GRCh38
rs748219859 Chr.X: 35951766-35951766 on Build GRCh38
rs372309727 Chr.X: 35951850-35951850 on Build GRCh38
rs753573604 Chr.X: 35951859-35951859 on Build GRCh38

View Full Product Details