Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21188588-21188827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS7B
Entrez Gene ID
25979
Gene Name
dehydrogenase/reductase 7B
Gene Aliases
CGI-93, SDR32C1
UniGene
Hs.386989
Gene Chromosome Location
Chr.17: 21123369-21191972 on Build GRCh38
Transcripts
NM_015510.4 XM_005256595.4 XM_005256596.3 XM_011523786.2 XM_017024425.1 XM_017024426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5646685 c.691G>C p.E231Q Chr.17: 21188782-21188782 on Build GRCh38
5758000 c.620A>T p.Y207F Chr.17: 21188711-21188711 on Build GRCh38
248768 c.665G>A p.R222H Chr.17: 21188756-21188756 on Build GRCh38
6643355 c.700G>A p.V234I Chr.17: 21188791-21188791 on Build GRCh38
6643354 c.712G>A p.G238S Chr.17: 21188803-21188803 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751820251 Chr.17: 21188774-21188774 on Build GRCh38
rs565974903 Chr.17: 21188654-21188654 on Build GRCh38
rs909921091 Chr.17: 21188647-21188647 on Build GRCh38
rs370896452 Chr.17: 21188733-21188733 on Build GRCh38
rs200124826 Chr.17: 21188674-21188674 on Build GRCh38
rs979625469 Chr.17: 21188622-21188625 on Build GRCh38
rs938157931 Chr.17: 21188627-21188627 on Build GRCh38
rs201946422 Chr.17: 21188725-21188725 on Build GRCh38
rs113030470 Chr.17: 21188669-21188669 on Build GRCh38
rs765409007 Chr.17: 21188662-21188662 on Build GRCh38
rs776219075 Chr.17: 21188748-21188748 on Build GRCh38
rs571452442 Chr.17: 21188678-21188678 on Build GRCh38
rs1048538917 Chr.17: 21188798-21188799 on Build GRCh38
rs746391576 Chr.17: 21188685-21188685 on Build GRCh38
rs763164881 Chr.17: 21188764-21188764 on Build GRCh38
rs752891452 Chr.17: 21188667-21188667 on Build GRCh38
rs538971297 Chr.17: 21188687-21188687 on Build GRCh38
rs1050975713 Chr.17: 21188760-21188760 on Build GRCh38
rs780529889 Chr.17: 21188694-21188694 on Build GRCh38
rs781355838 Chr.17: 21188683-21188683 on Build GRCh38
rs764039935 Chr.17: 21188766-21188766 on Build GRCh38
rs557704274 Chr.17: 21188720-21188720 on Build GRCh38
rs925443920 Chr.17: 21188628-21188628 on Build GRCh38
rs926653400 Chr.17: 21188696-21188696 on Build GRCh38
rs373037825 Chr.17: 21188670-21188670 on Build GRCh38
rs376241779 Chr.17: 21188727-21188727 on Build GRCh38
rs750847780 Chr.17: 21188791-21188791 on Build GRCh38
rs577733383 Chr.17: 21188790-21188790 on Build GRCh38
rs752656764 Chr.17: 21188761-21188761 on Build GRCh38
rs560838545 Chr.17: 21188629-21188629 on Build GRCh38
rs756450457 Chr.17: 21188803-21188803 on Build GRCh38
rs760193362 Chr.17: 21188728-21188728 on Build GRCh38
rs769405006 Chr.17: 21188765-21188766 on Build GRCh38
rs757357586 Chr.17: 21188784-21188784 on Build GRCh38
rs747710075 Chr.17: 21188711-21188711 on Build GRCh38
rs949378115 Chr.17: 21188675-21188675 on Build GRCh38
rs748031839 Chr.17: 21188676-21188679 on Build GRCh38
rs73307610 Chr.17: 21188618-21188618 on Build GRCh38
rs765323120 Chr.17: 21188756-21188756 on Build GRCh38
rs933925814 Chr.17: 21188707-21188707 on Build GRCh38
rs759397209 Chr.17: 21188755-21188755 on Build GRCh38

View Full Product Details