Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3872443-3872716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DFFB
Entrez Gene ID
1677
Gene Name
DNA fragmentation factor subunit beta
Gene Aliases
CAD, CPAN, DFF-40, DFF2, DFF40
UniGene
Hs.133089
Gene Chromosome Location
Chr.1: 3857143-3885429 on Build GRCh38
Transcripts
NM_001282669.1 NM_001320132.1 NM_001320136.1 NM_004402.3 NR_104222.1 NR_135150.1 NR_135151.1 NR_135152.1 XM_017000498.1 XM_017000499.1 XM_017000500.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6250643 c.774G>T p.L258L Chr.1: 3872564-3872564 on Build GRCh38
6231416 c.701G>T p.S234I Chr.1: 3872491-3872491 on Build GRCh38
908982 c.682-1G>T p.? Chr.1: 3872471-3872471 on Build GRCh38
5380894 c.710C>T p.S237L Chr.1: 3872500-3872500 on Build GRCh38
4455377 c.751A>C p.I251L Chr.1: 3872541-3872541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774923879 Chr.1: 3872561-3872561 on Build GRCh38
rs141093368 Chr.1: 3872607-3872607 on Build GRCh38
rs543420498 Chr.1: 3872693-3872693 on Build GRCh38
rs549184938 Chr.1: 3872485-3872485 on Build GRCh38
rs79548119 Chr.1: 3872648-3872648 on Build GRCh38
rs754718547 Chr.1: 3872490-3872490 on Build GRCh38
rs779412170 Chr.1: 3872582-3872583 on Build GRCh38
rs903383818 Chr.1: 3872649-3872665 on Build GRCh38
rs200599112 Chr.1: 3872467-3872467 on Build GRCh38
rs746071644 Chr.1: 3872665-3872665 on Build GRCh38
rs150541762 Chr.1: 3872497-3872497 on Build GRCh38
rs751751055 Chr.1: 3872546-3872546 on Build GRCh38
rs557852213 Chr.1: 3872666-3872666 on Build GRCh38
rs146190593 Chr.1: 3872523-3872523 on Build GRCh38
rs769747817 Chr.1: 3872608-3872608 on Build GRCh38
rs756823907 Chr.1: 3872470-3872470 on Build GRCh38
rs762248280 Chr.1: 3872562-3872562 on Build GRCh38
rs547878165 Chr.1: 3872567-3872567 on Build GRCh38
rs201229465 Chr.1: 3872618-3872618 on Build GRCh38
rs368137137 Chr.1: 3872555-3872555 on Build GRCh38
rs910681363 Chr.1: 3872657-3872657 on Build GRCh38
rs749891559 Chr.1: 3872576-3872576 on Build GRCh38
rs766944727 Chr.1: 3872472-3872472 on Build GRCh38
rs11581401 Chr.1: 3872631-3872631 on Build GRCh38
rs1006859074 Chr.1: 3872487-3872487 on Build GRCh38
rs778978825 Chr.1: 3872494-3872494 on Build GRCh38
rs771150232 Chr.1: 3872498-3872498 on Build GRCh38
rs755711660 Chr.1: 3872481-3872481 on Build GRCh38
rs146528418 Chr.1: 3872603-3872603 on Build GRCh38
rs750086063 Chr.1: 3872475-3872475 on Build GRCh38
rs75402433 Chr.1: 3872647-3872647 on Build GRCh38
rs879180387 Chr.1: 3872664-3872664 on Build GRCh38
rs913900686 Chr.1: 3872651-3872651 on Build GRCh38
rs754809517 Chr.1: 3872591-3872591 on Build GRCh38
rs376733287 Chr.1: 3872588-3872588 on Build GRCh38
rs72848483 Chr.1: 3872659-3872659 on Build GRCh38
rs185631375 Chr.1: 3872587-3872587 on Build GRCh38
rs70940336 Chr.1: 3872630-3872631 on Build GRCh38
rs368286589 Chr.1: 3872601-3872601 on Build GRCh38
rs746125016 Chr.1: 3872517-3872517 on Build GRCh38
rs751994951 Chr.1: 3872466-3872466 on Build GRCh38
rs577537997 Chr.1: 3872682-3872682 on Build GRCh38
rs748731886 Chr.1: 3872613-3872614 on Build GRCh38
rs755603298 Chr.1: 3872583-3872583 on Build GRCh38
rs993744209 Chr.1: 3872676-3872676 on Build GRCh38
rs755710345 Chr.1: 3872478-3872478 on Build GRCh38
rs12750562 Chr.1: 3872630-3872630 on Build GRCh38
rs749160520 Chr.1: 3872489-3872489 on Build GRCh38
rs202183040 Chr.1: 3872614-3872615 on Build GRCh38
rs374758045 Chr.1: 3872484-3872484 on Build GRCh38
rs960435824 Chr.1: 3872558-3872558 on Build GRCh38
rs776804679 Chr.1: 3872510-3872510 on Build GRCh38
rs748118488 Chr.1: 3872602-3872602 on Build GRCh38

View Full Product Details