Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 116912022-116912251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPZA2
Entrez Gene ID
830
Gene Name
capping actin protein of muscle Z-line alpha subunit 2
Gene Aliases
CAPPA2, CAPZ
UniGene
Hs.446123
Gene Chromosome Location
Chr.7: 116862478-116919261 on Build GRCh38
Transcripts
NM_006136.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6175965 c.598_599GA>TT p.E200L Chr.7: 116912081-116912082 on Build GRCh38
1084420 c.631G>T p.D211Y Chr.7: 116912114-116912114 on Build GRCh38
4945468 c.622A>G p.S208G Chr.7: 116912105-116912105 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762133426 Chr.7: 116912192-116912192 on Build GRCh38
rs201208269 Chr.7: 116912080-116912080 on Build GRCh38
rs774410549 Chr.7: 116912189-116912189 on Build GRCh38
rs757906913 Chr.7: 116912123-116912123 on Build GRCh38
rs769914360 Chr.7: 116912161-116912161 on Build GRCh38
rs1001131181 Chr.7: 116912206-116912206 on Build GRCh38
rs902623698 Chr.7: 116912207-116912207 on Build GRCh38
rs776530729 Chr.7: 116912157-116912157 on Build GRCh38
rs775585737 Chr.7: 116912167-116912167 on Build GRCh38
rs911579708 Chr.7: 116912101-116912101 on Build GRCh38
rs112570095 Chr.7: 116912098-116912098 on Build GRCh38
rs757611246 Chr.7: 116912118-116912118 on Build GRCh38
rs974998842 Chr.7: 116912119-116912119 on Build GRCh38
rs777250338 Chr.7: 116912125-116912125 on Build GRCh38
rs747599961 Chr.7: 116912121-116912121 on Build GRCh38
rs763062526 Chr.7: 116912181-116912181 on Build GRCh38
rs750860791 Chr.7: 116912086-116912086 on Build GRCh38
rs764268964 Chr.7: 116912182-116912182 on Build GRCh38
rs946905443 Chr.7: 116912170-116912170 on Build GRCh38
rs140848019 Chr.7: 116912127-116912127 on Build GRCh38
rs745797296 Chr.7: 116912160-116912160 on Build GRCh38

View Full Product Details