Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28530654-28530927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXN1
Entrez Gene ID
8456
Gene Name
forkhead box N1
Gene Aliases
FKHL20, RONU, WHN
UniGene
Hs.58611
Gene Chromosome Location
Chr.17: 28506211-28538900 on Build GRCh38
Transcripts
NM_003593.2 XM_005258046.3 XM_011525358.1 XM_011525359.1 XM_011525362.1 XM_011525367.1 XM_011525368.2 XM_011525369.2 XM_011525370.2 XM_017025228.1 XM_017025229.1 XM_017025230.1 XM_017025231.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3515383 c.846G>A p.M282I Chr.17: 28530764-28530764 on Build GRCh38
3515384 c.884G>A p.S295N Chr.17: 28530802-28530802 on Build GRCh38
6309433 c.877C>T p.P293S Chr.17: 28530795-28530795 on Build GRCh38
6666407 c.885C>T p.S295S Chr.17: 28530803-28530803 on Build GRCh38
148220 c.852T>C p.L284L Chr.17: 28530770-28530770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374220152 Chr.17: 28530797-28530797 on Build GRCh38
rs891631381 Chr.17: 28530681-28530681 on Build GRCh38
rs373841005 Chr.17: 28530788-28530788 on Build GRCh38
rs962507926 Chr.17: 28530865-28530865 on Build GRCh38
rs1022000988 Chr.17: 28530873-28530873 on Build GRCh38
rs118009648 Chr.17: 28530720-28530720 on Build GRCh38
rs764771520 Chr.17: 28530719-28530719 on Build GRCh38
rs371766542 Chr.17: 28530791-28530791 on Build GRCh38
rs953417623 Chr.17: 28530891-28530891 on Build GRCh38
rs550812773 Chr.17: 28530905-28530905 on Build GRCh38
rs140230257 Chr.17: 28530808-28530808 on Build GRCh38
rs751337959 Chr.17: 28530863-28530863 on Build GRCh38
rs764968394 Chr.17: 28530838-28530838 on Build GRCh38
rs879025096 Chr.17: 28530690-28530690 on Build GRCh38
rs778366866 Chr.17: 28530707-28530707 on Build GRCh38
rs777694363 Chr.17: 28530856-28530856 on Build GRCh38
rs751461103 Chr.17: 28530702-28530719 on Build GRCh38
rs781430179 Chr.17: 28530726-28530726 on Build GRCh38
rs113979879 Chr.17: 28530750-28530750 on Build GRCh38
rs757263962 Chr.17: 28530867-28530867 on Build GRCh38
rs200898611 Chr.17: 28530823-28530823 on Build GRCh38
rs560693790 Chr.17: 28530678-28530678 on Build GRCh38
rs968574119 Chr.17: 28530888-28530888 on Build GRCh38
rs977221161 Chr.17: 28530899-28530899 on Build GRCh38
rs1007949432 Chr.17: 28530762-28530762 on Build GRCh38
rs774193463 Chr.17: 28530740-28530740 on Build GRCh38
rs781234322 Chr.17: 28530874-28530874 on Build GRCh38
rs775627005 Chr.17: 28530812-28530812 on Build GRCh38
rs1006349854 Chr.17: 28530821-28530821 on Build GRCh38
rs758276674 Chr.17: 28530851-28530851 on Build GRCh38
rs773011041 Chr.17: 28530729-28530729 on Build GRCh38
rs753397380 Chr.17: 28530804-28530804 on Build GRCh38
rs752571655 Chr.17: 28530715-28530715 on Build GRCh38
rs752258395 Chr.17: 28530848-28530848 on Build GRCh38
rs772859622 Chr.17: 28530677-28530677 on Build GRCh38
rs754676566 Chr.17: 28530700-28530700 on Build GRCh38
rs745868096 Chr.17: 28530737-28530737 on Build GRCh38
rs945851109 Chr.17: 28530718-28530718 on Build GRCh38
rs911143626 Chr.17: 28530907-28530907 on Build GRCh38
rs780366040 Chr.17: 28530895-28530895 on Build GRCh38
rs767234511 Chr.17: 28530776-28530776 on Build GRCh38
rs12449554 Chr.17: 28530770-28530770 on Build GRCh38
rs12451259 Chr.17: 28530719-28530719 on Build GRCh38
rs745980479 Chr.17: 28530877-28530877 on Build GRCh38
rs767958911 Chr.17: 28530803-28530803 on Build GRCh38
rs762295420 Chr.17: 28530794-28530794 on Build GRCh38

View Full Product Details