Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207578166-207578391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CR1
Entrez Gene ID
1378
Gene Name
complement component 3b/4b receptor 1 (Knops blood group)
Gene Aliases
C3BR, C4BR, CD35, KN
UniGene
Hs.334019
Gene Chromosome Location
Chr.1: 207496128-207641765 on Build GRCh38
Transcripts
NM_000573.3 NM_000651.4 XM_006711166.2 XM_011509205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4674539 c.3562C>T p.P1188S Chr.1: 207578179-207578179 on Build GRCh38
6635057 c.4910A>G p.E1637G Chr.1: 207578177-207578177 on Build GRCh38
6635058 c.3560A>G p.E1187G Chr.1: 207578177-207578177 on Build GRCh38
4518004 c.4931_4932CC>TT p.S1644F Chr.1: 207578198-207578199 on Build GRCh38
344948 c.4909G>C p.E1637Q Chr.1: 207578176-207578176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368034349 Chr.1: 207578246-207578246 on Build GRCh38
rs749866185 Chr.1: 207578363-207578363 on Build GRCh38
rs938115813 Chr.1: 207578291-207578291 on Build GRCh38
rs781728384 Chr.1: 207578229-207578229 on Build GRCh38
rs758432098 Chr.1: 207578219-207578219 on Build GRCh38
rs975803399 Chr.1: 207578232-207578232 on Build GRCh38
rs759817653 Chr.1: 207578212-207578212 on Build GRCh38
rs566014769 Chr.1: 207578256-207578256 on Build GRCh38
rs926803324 Chr.1: 207578287-207578287 on Build GRCh38
rs771845752 Chr.1: 207578253-207578253 on Build GRCh38
rs769506190 Chr.1: 207578237-207578237 on Build GRCh38
rs373835373 Chr.1: 207578218-207578218 on Build GRCh38
rs745511420 Chr.1: 207578230-207578230 on Build GRCh38
rs1017404949 Chr.1: 207578207-207578207 on Build GRCh38
rs116178033 Chr.1: 207578295-207578295 on Build GRCh38
rs752006917 Chr.1: 207578252-207578254 on Build GRCh38
rs752050097 Chr.1: 207578227-207578227 on Build GRCh38
rs576926415 Chr.1: 207578288-207578288 on Build GRCh38
rs757624356 Chr.1: 207578228-207578228 on Build GRCh38
rs748789547 Chr.1: 207578244-207578244 on Build GRCh38
rs774323262 Chr.1: 207578247-207578247 on Build GRCh38
rs557063086 Chr.1: 207578281-207578281 on Build GRCh38
rs778306277 Chr.1: 207578222-207578222 on Build GRCh38
rs914155169 Chr.1: 207578274-207578274 on Build GRCh38
rs896516310 Chr.1: 207578297-207578297 on Build GRCh38
rs779867900 Chr.1: 207578240-207578240 on Build GRCh38
rs552192337 Chr.1: 207578245-207578245 on Build GRCh38
rs946948760 Chr.1: 207578275-207578275 on Build GRCh38
rs955695623 Chr.1: 207578254-207578254 on Build GRCh38
rs535010562 Chr.1: 207578294-207578294 on Build GRCh38
rs988406040 Chr.1: 207578257-207578257 on Build GRCh38

View Full Product Details