Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 98905355-98905580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL4
Entrez Gene ID
26235
Gene Name
F-box and leucine rich repeat protein 4
Gene Aliases
FBL4, FBL5, MTDPS13
UniGene
Hs.536850
Gene Chromosome Location
Chr.6: 98868535-98948006 on Build GRCh38
Transcripts
NM_001278716.1 NM_012160.4 NR_103836.1 NR_103837.1 XM_005266930.2 XM_011535748.2 XM_017010726.1 XM_017010727.1 XM_017010728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4457503 c.1048C>T p.L350F Chr.6: 98905481-98905481 on Build GRCh38
1737347 c.998A>G p.D333G Chr.6: 98905531-98905531 on Build GRCh38
3630715 c.1085C>T p.S362F Chr.6: 98905444-98905444 on Build GRCh38
6315006 c.1103+8C>A p.? Chr.6: 98905418-98905418 on Build GRCh38
5450302 c.954G>A p.L318L Chr.6: 98905575-98905575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199540184 Chr.6: 98905460-98905460 on Build GRCh38
rs780707718 Chr.6: 98905504-98905504 on Build GRCh38
rs759851318 Chr.6: 98905541-98905541 on Build GRCh38
rs368029297 Chr.6: 98905446-98905446 on Build GRCh38
rs764180868 Chr.6: 98905389-98905389 on Build GRCh38
rs200701723 Chr.6: 98905388-98905389 on Build GRCh38
rs182076681 Chr.6: 98905413-98905413 on Build GRCh38
rs759753713 Chr.6: 98905442-98905442 on Build GRCh38
rs377101732 Chr.6: 98905396-98905396 on Build GRCh38
rs367559138 Chr.6: 98905390-98905390 on Build GRCh38
rs1028748836 Chr.6: 98905469-98905469 on Build GRCh38
rs373555765 Chr.6: 98905491-98905491 on Build GRCh38
rs750183150 Chr.6: 98905394-98905394 on Build GRCh38
rs748620607 Chr.6: 98905398-98905398 on Build GRCh38
rs778318524 Chr.6: 98905520-98905520 on Build GRCh38
rs933217926 Chr.6: 98905539-98905539 on Build GRCh38
rs758028668 Chr.6: 98905395-98905395 on Build GRCh38
rs372544390 Chr.6: 98905456-98905456 on Build GRCh38
rs374114762 Chr.6: 98905414-98905414 on Build GRCh38
rs747657121 Chr.6: 98905509-98905509 on Build GRCh38
rs772370168 Chr.6: 98905547-98905547 on Build GRCh38
rs762333324 Chr.6: 98905507-98905508 on Build GRCh38
rs984719896 Chr.6: 98905507-98905507 on Build GRCh38
rs1045218869 Chr.6: 98905392-98905392 on Build GRCh38
rs778583873 Chr.6: 98905393-98905393 on Build GRCh38
rs761676733 Chr.6: 98905458-98905458 on Build GRCh38
rs749668327 Chr.6: 98905538-98905538 on Build GRCh38
rs774697942 Chr.6: 98905426-98905426 on Build GRCh38
rs150436385 Chr.6: 98905459-98905459 on Build GRCh38
rs6935315 Chr.6: 98905396-98905396 on Build GRCh38
rs758201051 Chr.6: 98905462-98905462 on Build GRCh38
rs771325864 Chr.6: 98905419-98905419 on Build GRCh38
rs762795927 Chr.6: 98905395-98905396 on Build GRCh38
rs745762480 Chr.6: 98905389-98905389 on Build GRCh38
rs189497833 Chr.6: 98905397-98905397 on Build GRCh38
rs779748858 Chr.6: 98905473-98905473 on Build GRCh38
rs775198265 Chr.6: 98905396-98905397 on Build GRCh38
rs186727069 Chr.6: 98905492-98905492 on Build GRCh38
rs754570800 Chr.6: 98905502-98905502 on Build GRCh38
rs770241351 Chr.6: 98905518-98905518 on Build GRCh38
rs368886622 Chr.6: 98905540-98905540 on Build GRCh38
rs984621399 Chr.6: 98905448-98905448 on Build GRCh38
rs559360649 Chr.6: 98905391-98905391 on Build GRCh38
rs61744041 Chr.6: 98905551-98905551 on Build GRCh38

View Full Product Details