Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 33389281-33389506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT10
Entrez Gene ID
84750
Gene Name
fucosyltransferase 10
Gene Aliases
FUCTX
UniGene
Hs.458713
Gene Chromosome Location
Chr.8: 33308102-33473492 on Build GRCh38
Transcripts
NM_032664.3 XM_011544676.2 XM_011544677.2 XM_011544678.2 XM_011544679.2 XM_011544680.2 XM_011544681.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4876940 c.664G>A p.V222I Chr.8: 33389325-33389325 on Build GRCh38
7183683 c.601G>A p.D201N Chr.8: 33389388-33389388 on Build GRCh38
3663742 c.556C>T p.R186* Chr.8: 33389433-33389433 on Build GRCh38
3929541 c.773A>G p.N258S Chr.8: 33389402-33389402 on Build GRCh38
6112965 c.743G>A p.R248Q Chr.8: 33389432-33389432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746005974 Chr.8: 33389368-33389368 on Build GRCh38
rs555345598 Chr.8: 33389481-33389481 on Build GRCh38
rs112717933 Chr.8: 33389320-33389320 on Build GRCh38
rs762803542 Chr.8: 33389394-33389394 on Build GRCh38
rs199683198 Chr.8: 33389383-33389383 on Build GRCh38
rs768827199 Chr.8: 33389447-33389447 on Build GRCh38
rs747557643 Chr.8: 33389335-33389335 on Build GRCh38
rs912944910 Chr.8: 33389411-33389411 on Build GRCh38
rs370808433 Chr.8: 33389461-33389461 on Build GRCh38
rs202210157 Chr.8: 33389405-33389405 on Build GRCh38
rs986304110 Chr.8: 33389415-33389415 on Build GRCh38
rs774094122 Chr.8: 33389460-33389460 on Build GRCh38
rs376361105 Chr.8: 33389454-33389454 on Build GRCh38
rs373622759 Chr.8: 33389464-33389464 on Build GRCh38
rs763714816 Chr.8: 33389470-33389470 on Build GRCh38
rs200415562 Chr.8: 33389479-33389479 on Build GRCh38
rs892199469 Chr.8: 33389341-33389341 on Build GRCh38
rs945836414 Chr.8: 33389423-33389423 on Build GRCh38
rs137929632 Chr.8: 33389396-33389396 on Build GRCh38
rs771351372 Chr.8: 33389356-33389356 on Build GRCh38
rs367637252 Chr.8: 33389480-33389480 on Build GRCh38
rs149448636 Chr.8: 33389432-33389432 on Build GRCh38
rs752208320 Chr.8: 33389463-33389463 on Build GRCh38
rs774998313 Chr.8: 33389360-33389360 on Build GRCh38
rs777417648 Chr.8: 33389422-33389422 on Build GRCh38
rs537662597 Chr.8: 33389400-33389400 on Build GRCh38
rs749608612 Chr.8: 33389343-33389343 on Build GRCh38
rs778105135 Chr.8: 33389411-33389412 on Build GRCh38
rs904207537 Chr.8: 33389451-33389451 on Build GRCh38
rs555909648 Chr.8: 33389449-33389449 on Build GRCh38
rs747040014 Chr.8: 33389446-33389446 on Build GRCh38
rs764316017 Chr.8: 33389385-33389385 on Build GRCh38
rs16880853 Chr.8: 33389373-33389373 on Build GRCh38
rs572787845 Chr.8: 33389455-33389455 on Build GRCh38
rs1016284356 Chr.8: 33389318-33389318 on Build GRCh38
rs747769477 Chr.8: 33389305-33389305 on Build GRCh38
rs767047032 Chr.8: 33389397-33389397 on Build GRCh38
rs774984724 Chr.8: 33389382-33389382 on Build GRCh38
rs772883445 Chr.8: 33389391-33389391 on Build GRCh38
rs369185532 Chr.8: 33389440-33389440 on Build GRCh38
rs540242281 Chr.8: 33389458-33389458 on Build GRCh38
rs752155322 Chr.8: 33389401-33389401 on Build GRCh38
rs145932857 Chr.8: 33389433-33389433 on Build GRCh38
rs753509956 Chr.8: 33389476-33389476 on Build GRCh38
rs748431671 Chr.8: 33389486-33389486 on Build GRCh38
rs143890078 Chr.8: 33389483-33389483 on Build GRCh38
rs567981527 Chr.8: 33389311-33389311 on Build GRCh38
rs772655040 Chr.8: 33389441-33389441 on Build GRCh38
rs775722431 Chr.8: 33389443-33389443 on Build GRCh38
rs772659526 Chr.8: 33389363-33389364 on Build GRCh38
rs140205664 Chr.8: 33389478-33389478 on Build GRCh38
rs778021782 Chr.8: 33389333-33389333 on Build GRCh38

View Full Product Details