Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 153887567-153887840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP6
Entrez Gene ID
1804
Gene Name
dipeptidyl peptidase like 6
Gene Aliases
DPL1, DPPX, MRD33, VF2
UniGene
Hs.490684
Gene Chromosome Location
Chr.7: 153747986-154894290 on Build GRCh38
Transcripts
NM_001039350.2 XM_017011808.1 XM_017011809.1 XM_017011810.1 XM_017011811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6109503 c.29C>T p.A10V Chr.7: 153887712-153887712 on Build GRCh38
354576 c.50T>C p.M17T Chr.7: 153887733-153887733 on Build GRCh38
5679824 c.38A>C p.Q13P Chr.7: 153887721-153887721 on Build GRCh38
3304862 c.36G>A p.M12I Chr.7: 153887719-153887719 on Build GRCh38
5657533 c.17T>G p.M6R Chr.7: 153887700-153887700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748265102 Chr.7: 153887695-153887695 on Build GRCh38
rs939021106 Chr.7: 153887804-153887804 on Build GRCh38
rs1057123177 Chr.7: 153887628-153887628 on Build GRCh38
rs1027468086 Chr.7: 153887765-153887765 on Build GRCh38
rs188850191 Chr.7: 153887673-153887673 on Build GRCh38
rs763832891 Chr.7: 153887772-153887772 on Build GRCh38
rs761086513 Chr.7: 153887767-153887767 on Build GRCh38
rs770225949 Chr.7: 153887763-153887763 on Build GRCh38
rs75677723 Chr.7: 153887698-153887698 on Build GRCh38
rs751101009 Chr.7: 153887731-153887731 on Build GRCh38
rs376727848 Chr.7: 153887676-153887676 on Build GRCh38
rs766860746 Chr.7: 153887699-153887699 on Build GRCh38
rs368684179 Chr.7: 153887748-153887748 on Build GRCh38
rs560934140 Chr.7: 153887740-153887740 on Build GRCh38
rs763709977 Chr.7: 153887728-153887728 on Build GRCh38
rs374452583 Chr.7: 153887747-153887747 on Build GRCh38
rs761134135 Chr.7: 153887643-153887643 on Build GRCh38
rs747090629 Chr.7: 153887751-153887751 on Build GRCh38
rs776929713 Chr.7: 153887710-153887710 on Build GRCh38
rs751154144 Chr.7: 153887776-153887776 on Build GRCh38
rs768016526 Chr.7: 153887729-153887730 on Build GRCh38
rs752416547 Chr.7: 153887725-153887725 on Build GRCh38
rs879170738 Chr.7: 153887646-153887646 on Build GRCh38
rs781150778 Chr.7: 153887734-153887734 on Build GRCh38
rs374588871 Chr.7: 153887821-153887821 on Build GRCh38
rs762340157 Chr.7: 153887716-153887724 on Build GRCh38
rs558844491 Chr.7: 153887654-153887654 on Build GRCh38
rs777175723 Chr.7: 153887760-153887760 on Build GRCh38
rs745670100 Chr.7: 153887688-153887688 on Build GRCh38
rs764849380 Chr.7: 153887724-153887724 on Build GRCh38
rs907595783 Chr.7: 153887780-153887780 on Build GRCh38
rs759916108 Chr.7: 153887717-153887717 on Build GRCh38
rs772166530 Chr.7: 153887696-153887696 on Build GRCh38
rs764940057 Chr.7: 153887661-153887661 on Build GRCh38
rs577074839 Chr.7: 153887663-153887663 on Build GRCh38
rs762751898 Chr.7: 153887768-153887768 on Build GRCh38
rs563257661 Chr.7: 153887617-153887617 on Build GRCh38
rs1011858807 Chr.7: 153887733-153887733 on Build GRCh38
rs768798277 Chr.7: 153887690-153887690 on Build GRCh38
rs371202831 Chr.7: 153887732-153887732 on Build GRCh38
rs1036118328 Chr.7: 153887609-153887609 on Build GRCh38
rs755955933 Chr.7: 153887741-153887741 on Build GRCh38
rs374919854 Chr.7: 153887650-153887650 on Build GRCh38
rs754648778 Chr.7: 153887660-153887660 on Build GRCh38
rs771469463 Chr.7: 153887759-153887759 on Build GRCh38
rs774607192 Chr.7: 153887694-153887694 on Build GRCh38
rs759911333 Chr.7: 153887761-153887761 on Build GRCh38
rs754176222 Chr.7: 153887655-153887655 on Build GRCh38
rs541759832 Chr.7: 153887607-153887607 on Build GRCh38
rs775658655 Chr.7: 153887767-153887767 on Build GRCh38
rs906914802 Chr.7: 153887624-153887624 on Build GRCh38
rs572133583 Chr.7: 153887679-153887679 on Build GRCh38
rs773757572 Chr.7: 153887766-153887767 on Build GRCh38
rs569697342 Chr.7: 153887600-153887600 on Build GRCh38
rs553289315 Chr.7: 153887674-153887674 on Build GRCh38
rs895359371 Chr.7: 153887644-153887644 on Build GRCh38
rs779670481 Chr.7: 153887745-153887745 on Build GRCh38

View Full Product Details