Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27369237-27369508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCN3
Entrez Gene ID
8547
Gene Name
ficolin 3
Gene Aliases
FCNH, HAKA1
UniGene
Hs.333383
Gene Chromosome Location
Chr.1: 27369110-27374824 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
907868 c.779G>A p.R260Q Chr.1: 27369357-27369357 on Build GRCh38
5797046 c.823C>T p.H275Y Chr.1: 27369313-27369313 on Build GRCh38
6484746 c.697A>T p.T233S Chr.1: 27369439-27369439 on Build GRCh38
5894204 c.683G>A p.G228E Chr.1: 27369453-27369453 on Build GRCh38
6126209 c.779G>T p.R260L Chr.1: 27369357-27369357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756190859 Chr.1: 27369402-27369402 on Build GRCh38
rs763335663 Chr.1: 27369457-27369457 on Build GRCh38
rs771519454 Chr.1: 27369463-27369463 on Build GRCh38
rs751750968 Chr.1: 27369269-27369269 on Build GRCh38
rs1046045160 Chr.1: 27369428-27369428 on Build GRCh38
rs761471363 Chr.1: 27369345-27369345 on Build GRCh38
rs746674895 Chr.1: 27369339-27369339 on Build GRCh38
rs767839461 Chr.1: 27369381-27369381 on Build GRCh38
rs774568591 Chr.1: 27369477-27369477 on Build GRCh38
rs544521222 Chr.1: 27369340-27369340 on Build GRCh38
rs773813058 Chr.1: 27369449-27369449 on Build GRCh38
rs201654480 Chr.1: 27369300-27369300 on Build GRCh38
rs143244488 Chr.1: 27369357-27369357 on Build GRCh38
rs745599639 Chr.1: 27369316-27369316 on Build GRCh38
rs781103320 Chr.1: 27369281-27369281 on Build GRCh38
rs1018094906 Chr.1: 27369385-27369385 on Build GRCh38
rs374013809 Chr.1: 27369425-27369425 on Build GRCh38
rs746876244 Chr.1: 27369431-27369431 on Build GRCh38
rs751993679 Chr.1: 27369362-27369362 on Build GRCh38
rs555099259 Chr.1: 27369382-27369382 on Build GRCh38
rs765768419 Chr.1: 27369346-27369346 on Build GRCh38
rs760012512 Chr.1: 27369484-27369484 on Build GRCh38
rs55863154 Chr.1: 27369276-27369276 on Build GRCh38
rs76041454 Chr.1: 27369317-27369317 on Build GRCh38
rs773222505 Chr.1: 27369283-27369283 on Build GRCh38
rs745832502 Chr.1: 27369422-27369422 on Build GRCh38
rs143220107 Chr.1: 27369430-27369430 on Build GRCh38
rs947815585 Chr.1: 27369455-27369455 on Build GRCh38
rs138304758 Chr.1: 27369374-27369374 on Build GRCh38
rs768187268 Chr.1: 27369434-27369434 on Build GRCh38
rs1028104779 Chr.1: 27369333-27369333 on Build GRCh38
rs575296520 Chr.1: 27369329-27369329 on Build GRCh38
rs141468956 Chr.1: 27369358-27369358 on Build GRCh38
rs28385652 Chr.1: 27369386-27369386 on Build GRCh38
rs369799164 Chr.1: 27369262-27369262 on Build GRCh38
rs914913323 Chr.1: 27369446-27369446 on Build GRCh38
rs778768902 Chr.1: 27369421-27369421 on Build GRCh38
rs989156066 Chr.1: 27369435-27369435 on Build GRCh38
rs769451051 Chr.1: 27369448-27369448 on Build GRCh38
rs953487707 Chr.1: 27369334-27369334 on Build GRCh38
rs34295487 Chr.1: 27369286-27369286 on Build GRCh38
rs1003725297 Chr.1: 27369265-27369265 on Build GRCh38
rs1045183385 Chr.1: 27369465-27369465 on Build GRCh38
rs34404009 Chr.1: 27369471-27369472 on Build GRCh38
rs553026191 Chr.1: 27369261-27369261 on Build GRCh38
rs776423358 Chr.1: 27369438-27369438 on Build GRCh38
rs748278677 Chr.1: 27369282-27369282 on Build GRCh38
rs747603248 Chr.1: 27369445-27369445 on Build GRCh38
rs775989626 Chr.1: 27369342-27369342 on Build GRCh38

View Full Product Details