Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75370020-75370273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPS2
Entrez Gene ID
84698
Gene Name
calcyphosine 2
Gene Aliases
UG0636c06
UniGene
Hs.733835
Gene Chromosome Location
Chr.12: 75275979-75390941 on Build GRCh38
Transcripts
NM_001286548.1 NM_001286549.1 XM_005269192.1 XM_011538888.1 XM_011538889.2 XM_011538892.1 XM_017020034.1 XM_017020035.1 XM_017020037.1 XM_017020038.1 XM_017020043.1 XM_017020044.1 XM_017020045.1 NM_001304964.1 NM_152779.3

Gene Symbol
GLIPR1L1
Entrez Gene ID
256710
Gene Name
GLI pathogenesis related 1 like 1
Gene Aliases
ALKN2972, PRO7434
UniGene
Hs.567788
Gene Chromosome Location
Chr.12: 75331834-75370390 on Build GRCh38
Transcripts
NM_001286548.1 NM_001286549.1 XM_005269192.1 XM_011538888.1 XM_011538889.2 XM_011538892.1 XM_017020034.1 XM_017020035.1 XM_017020037.1 XM_017020038.1 XM_017020043.1 XM_017020044.1 XM_017020045.1 NM_001304964.1 NM_152779.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2102408 c.622C>T p.L208L Chr.12: 75370096-75370096 on Build GRCh38
2102409 c.631A>G p.T211A Chr.12: 75370105-75370105 on Build GRCh38
5158895 c.660G>A p.T220T Chr.12: 75370107-75370107 on Build GRCh38
4211263 c.658A>G p.T220A Chr.12: 75370105-75370105 on Build GRCh38
6462955 c.611-5G>C p.? Chr.12: 75370080-75370080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750054991 Chr.12: 75370138-75370138 on Build GRCh38
rs529316132 Chr.12: 75370182-75370182 on Build GRCh38
rs781725841 Chr.12: 75370179-75370179 on Build GRCh38
rs1005624379 Chr.12: 75370062-75370062 on Build GRCh38
rs768648969 Chr.12: 75370129-75370129 on Build GRCh38
rs766648852 Chr.12: 75370150-75370150 on Build GRCh38
rs774756061 Chr.12: 75370210-75370214 on Build GRCh38
rs758783526 Chr.12: 75370069-75370069 on Build GRCh38
rs746821170 Chr.12: 75370228-75370228 on Build GRCh38
rs541396838 Chr.12: 75370196-75370196 on Build GRCh38
rs373880695 Chr.12: 75370082-75370082 on Build GRCh38
rs373471861 Chr.12: 75370171-75370171 on Build GRCh38
rs936457676 Chr.12: 75370195-75370195 on Build GRCh38
rs760513951 Chr.12: 75370121-75370121 on Build GRCh38
rs779652305 Chr.12: 75370201-75370201 on Build GRCh38
rs745613464 Chr.12: 75370112-75370112 on Build GRCh38
rs750745297 Chr.12: 75370057-75370057 on Build GRCh38
rs572445989 Chr.12: 75370153-75370153 on Build GRCh38
rs771237252 Chr.12: 75370051-75370052 on Build GRCh38
rs1016048028 Chr.12: 75370063-75370063 on Build GRCh38
rs563818649 Chr.12: 75370164-75370164 on Build GRCh38
rs749513253 Chr.12: 75370190-75370190 on Build GRCh38
rs778175786 Chr.12: 75370111-75370111 on Build GRCh38
rs935026159 Chr.12: 75370208-75370209 on Build GRCh38
rs756287444 Chr.12: 75370186-75370186 on Build GRCh38
rs775418934 Chr.12: 75370115-75370115 on Build GRCh38
rs148073028 Chr.12: 75370107-75370107 on Build GRCh38
rs775934608 Chr.12: 75370130-75370130 on Build GRCh38
rs375572537 Chr.12: 75370080-75370080 on Build GRCh38
rs970259313 Chr.12: 75370178-75370178 on Build GRCh38
rs771333853 Chr.12: 75370193-75370193 on Build GRCh38
rs916306625 Chr.12: 75370171-75370171 on Build GRCh38
rs533449405 Chr.12: 75370106-75370106 on Build GRCh38
rs755383804 Chr.12: 75370160-75370160 on Build GRCh38
rs770077159 Chr.12: 75370109-75370109 on Build GRCh38
rs148521297 Chr.12: 75370159-75370159 on Build GRCh38
rs764819843 Chr.12: 75370137-75370137 on Build GRCh38
rs755576011 Chr.12: 75370105-75370105 on Build GRCh38
rs200172257 Chr.12: 75370142-75370142 on Build GRCh38
rs559463316 Chr.12: 75370188-75370188 on Build GRCh38
rs780446030 Chr.12: 75370073-75370073 on Build GRCh38
rs764872848 Chr.12: 75370055-75370055 on Build GRCh38

View Full Product Details