Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11346488-11346739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC159
Entrez Gene ID
126075
Gene Name
coiled-coil domain containing 159
Gene Aliases
-
UniGene
Hs.631636
Gene Chromosome Location
Chr.19: 11346340-11354951 on Build GRCh38
Transcripts
NM_001080503.2 XM_006722643.1 XM_011527678.2 XM_017026255.1 XM_017026256.1 XM_017026258.1 XM_017026259.1 XM_017026260.1 XM_017026261.1 XM_017026262.1 XM_017026263.1 XM_017026264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5036721 c.21+4T>A p.? Chr.19: 11346631-11346631 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs957310073 Chr.19: 11346698-11346698 on Build GRCh38
rs1001271723 Chr.19: 11346682-11346682 on Build GRCh38
rs925280377 Chr.19: 11346582-11346582 on Build GRCh38
rs573470376 Chr.19: 11346699-11346699 on Build GRCh38
rs911303163 Chr.19: 11346596-11346596 on Build GRCh38
rs8105115 Chr.19: 11346702-11346702 on Build GRCh38
rs978770634 Chr.19: 11346571-11346571 on Build GRCh38
rs555253746 Chr.19: 11346685-11346685 on Build GRCh38
rs768373205 Chr.19: 11346599-11346599 on Build GRCh38
rs540129611 Chr.19: 11346684-11346684 on Build GRCh38
rs879406843 Chr.19: 11346618-11346618 on Build GRCh38
rs572397334 Chr.19: 11346573-11346573 on Build GRCh38
rs538792560 Chr.19: 11346533-11346533 on Build GRCh38
rs923129517 Chr.19: 11346713-11346713 on Build GRCh38
rs761709546 Chr.19: 11346677-11346677 on Build GRCh38
rs571729939 Chr.19: 11346528-11346528 on Build GRCh38
rs1013366150 Chr.19: 11346716-11346717 on Build GRCh38
rs369389941 Chr.19: 11346679-11346679 on Build GRCh38
rs958289109 Chr.19: 11346591-11346591 on Build GRCh38
rs894883967 Chr.19: 11346716-11346716 on Build GRCh38
rs554117030 Chr.19: 11346535-11346535 on Build GRCh38
rs944158089 Chr.19: 11346613-11346613 on Build GRCh38
rs1034061020 Chr.19: 11346686-11346686 on Build GRCh38
rs879198574 Chr.19: 11346711-11346711 on Build GRCh38
rs985521588 Chr.19: 11346593-11346593 on Build GRCh38
rs776479629 Chr.19: 11346651-11346651 on Build GRCh38
rs967339782 Chr.19: 11346706-11346706 on Build GRCh38
rs929215366 Chr.19: 11346688-11346688 on Build GRCh38

View Full Product Details