Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9938054-9938281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRELD1
Entrez Gene ID
78987
Gene Name
cysteine rich with EGF like domains 1
Gene Aliases
AVSD2, CIRRIN
UniGene
Hs.9383
Gene Chromosome Location
Chr.3: 9933788-9945413 on Build GRCh38
Transcripts
NM_001031717.3 NM_001077415.2 NM_015513.4 XM_011534108.1 XM_017007175.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1049358 c.460+1G>A p.? Chr.3: 9938107-9938107 on Build GRCh38
1249117 c.432C>T p.P144P Chr.3: 9938078-9938078 on Build GRCh38
1249118 c.432C>T p.P144P Chr.3: 9938078-9938078 on Build GRCh38
1594174 c.460+1G>A p.? Chr.3: 9938107-9938107 on Build GRCh38
6360513 c.455G>T p.C152F Chr.3: 9938101-9938101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970218876 Chr.3: 9938098-9938098 on Build GRCh38
rs931804745 Chr.3: 9938213-9938213 on Build GRCh38
rs780167927 Chr.3: 9938107-9938107 on Build GRCh38
rs373557385 Chr.3: 9938090-9938090 on Build GRCh38
rs1050134256 Chr.3: 9938195-9938195 on Build GRCh38
rs925136162 Chr.3: 9938228-9938228 on Build GRCh38
rs765996826 Chr.3: 9938147-9938147 on Build GRCh38
rs376349420 Chr.3: 9938105-9938105 on Build GRCh38
rs764703613 Chr.3: 9938138-9938138 on Build GRCh38
rs1041532498 Chr.3: 9938221-9938221 on Build GRCh38
rs966140248 Chr.3: 9938184-9938184 on Build GRCh38
rs975984863 Chr.3: 9938200-9938200 on Build GRCh38
rs761275781 Chr.3: 9938074-9938074 on Build GRCh38
rs747855475 Chr.3: 9938091-9938091 on Build GRCh38
rs376939409 Chr.3: 9938139-9938139 on Build GRCh38
rs907538434 Chr.3: 9938165-9938165 on Build GRCh38
rs1014507351 Chr.3: 9938080-9938080 on Build GRCh38
rs377654613 Chr.3: 9938093-9938093 on Build GRCh38
rs1043597852 Chr.3: 9938224-9938224 on Build GRCh38
rs200737884 Chr.3: 9938079-9938079 on Build GRCh38
rs920234388 Chr.3: 9938167-9938167 on Build GRCh38
rs921851984 Chr.3: 9938206-9938206 on Build GRCh38
rs767025754 Chr.3: 9938158-9938158 on Build GRCh38
rs777003297 Chr.3: 9938143-9938143 on Build GRCh38
rs754616906 Chr.3: 9938087-9938087 on Build GRCh38
rs201495314 Chr.3: 9938103-9938103 on Build GRCh38
rs754706628 Chr.3: 9938153-9938153 on Build GRCh38
rs751041958 Chr.3: 9938148-9938148 on Build GRCh38
rs185522163 Chr.3: 9938177-9938177 on Build GRCh38
rs936063857 Chr.3: 9938233-9938233 on Build GRCh38
rs750003238 Chr.3: 9938078-9938078 on Build GRCh38
rs911634389 Chr.3: 9938196-9938196 on Build GRCh38
rs762613034 Chr.3: 9938144-9938144 on Build GRCh38
rs760928235 Chr.3: 9938132-9938132 on Build GRCh38
rs775946538 Chr.3: 9938124-9938124 on Build GRCh38
rs370830167 Chr.3: 9938096-9938096 on Build GRCh38
rs772244142 Chr.3: 9938120-9938120 on Build GRCh38
rs746095271 Chr.3: 9938106-9938106 on Build GRCh38
rs1002124262 Chr.3: 9938102-9938102 on Build GRCh38
rs751240157 Chr.3: 9938082-9938082 on Build GRCh38

View Full Product Details