Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19183201-19183436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLTCL1
Entrez Gene ID
8218
Gene Name
clathrin heavy chain like 1
Gene Aliases
CHC22, CLH22, CLTCL, CLTD
UniGene
Hs.368266
Gene Chromosome Location
Chr.22: 19179473-19291716 on Build GRCh38
Transcripts
NM_001835.3 NM_007098.3 XM_011530401.2 XM_017028953.1 XM_017028954.1 XM_017028955.1 XM_017028956.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3424030 c.4783G>C p.A1595P Chr.22: 19183434-19183434 on Build GRCh38
2892089 c.4814A>G p.E1605G Chr.22: 19183403-19183403 on Build GRCh38
4297188 c.4814A>G p.E1605G Chr.22: 19183403-19183403 on Build GRCh38
3424029 c.4783G>C p.A1595P Chr.22: 19183434-19183434 on Build GRCh38
3379268 c.4815G>A p.E1605E Chr.22: 19183402-19183402 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782752175 Chr.22: 19183378-19183378 on Build GRCh38
rs35109488 Chr.22: 19183376-19183376 on Build GRCh38
rs115478161 Chr.22: 19183375-19183375 on Build GRCh38
rs922554685 Chr.22: 19183230-19183230 on Build GRCh38
rs1025680672 Chr.22: 19183416-19183416 on Build GRCh38
rs759640744 Chr.22: 19183383-19183383 on Build GRCh38
rs994403917 Chr.22: 19183397-19183397 on Build GRCh38
rs781902251 Chr.22: 19183399-19183399 on Build GRCh38
rs113585066 Chr.22: 19183318-19183318 on Build GRCh38
rs782014565 Chr.22: 19183362-19183362 on Build GRCh38
rs573203884 Chr.22: 19183381-19183381 on Build GRCh38
rs782238358 Chr.22: 19183359-19183359 on Build GRCh38
rs368354741 Chr.22: 19183393-19183393 on Build GRCh38
rs782709145 Chr.22: 19183361-19183361 on Build GRCh38
rs771076182 Chr.22: 19183275-19183275 on Build GRCh38
rs781818684 Chr.22: 19183315-19183315 on Build GRCh38
rs977079186 Chr.22: 19183263-19183263 on Build GRCh38
rs782284164 Chr.22: 19183371-19183371 on Build GRCh38
rs377049848 Chr.22: 19183380-19183380 on Build GRCh38
rs782813231 Chr.22: 19183390-19183390 on Build GRCh38
rs545321400 Chr.22: 19183415-19183415 on Build GRCh38
rs945961513 Chr.22: 19183232-19183232 on Build GRCh38
rs373832647 Chr.22: 19183373-19183373 on Build GRCh38
rs782435776 Chr.22: 19183406-19183406 on Build GRCh38
rs573578230 Chr.22: 19183319-19183319 on Build GRCh38
rs925606086 Chr.22: 19183284-19183284 on Build GRCh38

View Full Product Details