Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35747792-35748031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS2L2
Entrez Gene ID
246176
Gene Name
growth arrest specific 2 like 2
Gene Aliases
GAR17
UniGene
Hs.591194
Gene Chromosome Location
Chr.17: 35744511-35752878 on Build GRCh38
Transcripts
NM_139285.3

SNP Detail

SNP ID Chromosome Location
rs782255408 Chr.17: 35747814-35747814 on Build GRCh38
rs898143569 Chr.17: 35748010-35748010 on Build GRCh38
rs782287283 Chr.17: 35747960-35747960 on Build GRCh38
rs782624210 Chr.17: 35747875-35747875 on Build GRCh38
rs782359008 Chr.17: 35747980-35747980 on Build GRCh38
rs782650477 Chr.17: 35747911-35747911 on Build GRCh38
rs782529922 Chr.17: 35747873-35747873 on Build GRCh38
rs782794658 Chr.17: 35747838-35747838 on Build GRCh38
rs587724852 Chr.17: 35747823-35747823 on Build GRCh38
rs140842796 Chr.17: 35747864-35747864 on Build GRCh38
rs782738507 Chr.17: 35747944-35747944 on Build GRCh38
rs937820006 Chr.17: 35747970-35747970 on Build GRCh38
rs782445806 Chr.17: 35747957-35747957 on Build GRCh38
rs201670008 Chr.17: 35747927-35747927 on Build GRCh38
rs781941592 Chr.17: 35747984-35747984 on Build GRCh38
rs781888671 Chr.17: 35747872-35747872 on Build GRCh38
rs587766132 Chr.17: 35748011-35748011 on Build GRCh38
rs782390186 Chr.17: 35747998-35747998 on Build GRCh38
rs782674519 Chr.17: 35747813-35747813 on Build GRCh38
rs372666616 Chr.17: 35747986-35747986 on Build GRCh38
rs782227203 Chr.17: 35747972-35747972 on Build GRCh38
rs782386293 Chr.17: 35747920-35747920 on Build GRCh38
rs912636599 Chr.17: 35747819-35747819 on Build GRCh38
rs781925735 Chr.17: 35747832-35747832 on Build GRCh38
rs986884387 Chr.17: 35747815-35747815 on Build GRCh38
rs202192424 Chr.17: 35747921-35747921 on Build GRCh38
rs782688271 Chr.17: 35747959-35747959 on Build GRCh38
rs781925975 Chr.17: 35747928-35747928 on Build GRCh38
rs896240400 Chr.17: 35747976-35747976 on Build GRCh38
rs782008707 Chr.17: 35747846-35747846 on Build GRCh38
rs782059575 Chr.17: 35747985-35747985 on Build GRCh38
rs587627832 Chr.17: 35747939-35747939 on Build GRCh38
rs374053408 Chr.17: 35747973-35747973 on Build GRCh38
rs587754002 Chr.17: 35747866-35747866 on Build GRCh38
rs142989535 Chr.17: 35747886-35747886 on Build GRCh38
rs147920035 Chr.17: 35747938-35747938 on Build GRCh38
rs146777075 Chr.17: 35747930-35747930 on Build GRCh38
rs914534605 Chr.17: 35747903-35747903 on Build GRCh38
rs782323809 Chr.17: 35747830-35747830 on Build GRCh38
rs201121527 Chr.17: 35747890-35747890 on Build GRCh38
rs782454642 Chr.17: 35747865-35747865 on Build GRCh38
rs1015274777 Chr.17: 35748006-35748006 on Build GRCh38
rs587679089 Chr.17: 35748009-35748009 on Build GRCh38
rs782082920 Chr.17: 35747925-35747925 on Build GRCh38
rs147743370 Chr.17: 35747896-35747896 on Build GRCh38
rs782428597 Chr.17: 35747909-35747909 on Build GRCh38
rs782372216 Chr.17: 35747816-35747816 on Build GRCh38
rs782178910 Chr.17: 35747907-35747907 on Build GRCh38
rs371875125 Chr.17: 35747918-35747918 on Build GRCh38
rs782211317 Chr.17: 35747828-35747828 on Build GRCh38
rs782642508 Chr.17: 35747971-35747971 on Build GRCh38
rs782111847 Chr.17: 35747858-35747858 on Build GRCh38
rs781802273 Chr.17: 35747948-35747948 on Build GRCh38
rs200562123 Chr.17: 35747974-35747974 on Build GRCh38
rs782527270 Chr.17: 35747965-35747965 on Build GRCh38
rs377312376 Chr.17: 35748011-35748011 on Build GRCh38
rs145467681 Chr.17: 35747859-35747859 on Build GRCh38

View Full Product Details