Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 76346735-76346990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBS10
Entrez Gene ID
79738
Gene Name
Bardet-Biedl syndrome 10
Gene Aliases
C12orf58
UniGene
Hs.96322
Gene Chromosome Location
Chr.12: 76344486-76348442 on Build GRCh38
Transcripts
NM_024685.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
943531 c.1073C>T p.S358L Chr.12: 76346912-76346912 on Build GRCh38
4665253 c.1216C>A p.L406I Chr.12: 76346769-76346769 on Build GRCh38
943532 c.1027C>A p.R343R Chr.12: 76346958-76346958 on Build GRCh38
5212170 c.1139delG p.R380fs*5 Chr.12: 76346846-76346846 on Build GRCh38
6000294 c.1210C>T p.H404Y Chr.12: 76346775-76346775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756934245 Chr.12: 76346917-76346917 on Build GRCh38
rs751592378 Chr.12: 76346770-76346770 on Build GRCh38
rs947783482 Chr.12: 76346874-76346874 on Build GRCh38
rs370953693 Chr.12: 76346935-76346935 on Build GRCh38
rs747390360 Chr.12: 76346944-76346944 on Build GRCh38
rs727503818 Chr.12: 76346894-76346894 on Build GRCh38
rs371878955 Chr.12: 76346816-76346816 on Build GRCh38
rs727503819 Chr.12: 76346895-76346895 on Build GRCh38
rs775492103 Chr.12: 76346841-76346841 on Build GRCh38
rs786204573 Chr.12: 76346800-76346801 on Build GRCh38
rs938066133 Chr.12: 76346897-76346897 on Build GRCh38
rs774809155 Chr.12: 76346831-76346831 on Build GRCh38
rs867485840 Chr.12: 76346798-76346798 on Build GRCh38
rs202042386 Chr.12: 76346796-76346796 on Build GRCh38
rs747097629 Chr.12: 76346946-76346946 on Build GRCh38
rs920028641 Chr.12: 76346870-76346870 on Build GRCh38
rs550120113 Chr.12: 76346953-76346953 on Build GRCh38
rs1057517206 Chr.12: 76346794-76346795 on Build GRCh38
rs761897354 Chr.12: 76346951-76346951 on Build GRCh38
rs551803123 Chr.12: 76346911-76346911 on Build GRCh38
rs199474722 Chr.12: 76346783-76346783 on Build GRCh38
rs377164732 Chr.12: 76346818-76346818 on Build GRCh38
rs746334861 Chr.12: 76346832-76346832 on Build GRCh38
rs863224793 Chr.12: 76346800-76346800 on Build GRCh38
rs758194221 Chr.12: 76346767-76346767 on Build GRCh38
rs974452621 Chr.12: 76346788-76346788 on Build GRCh38
rs775926390 Chr.12: 76346945-76346945 on Build GRCh38
rs760673745 Chr.12: 76346845-76346845 on Build GRCh38
rs750164736 Chr.12: 76346765-76346765 on Build GRCh38
rs200605410 Chr.12: 76346949-76346949 on Build GRCh38
rs766405505 Chr.12: 76346769-76346769 on Build GRCh38
rs777358585 Chr.12: 76346817-76346817 on Build GRCh38
rs753604828 Chr.12: 76346862-76346863 on Build GRCh38
rs143903199 Chr.12: 76346797-76346797 on Build GRCh38
rs762785764 Chr.12: 76346860-76346860 on Build GRCh38
rs1057516623 Chr.12: 76346929-76346929 on Build GRCh38
rs530892122 Chr.12: 76346948-76346948 on Build GRCh38
rs764371181 Chr.12: 76346851-76346851 on Build GRCh38
rs759474648 Chr.12: 76346958-76346958 on Build GRCh38
rs587777837 Chr.12: 76346940-76346941 on Build GRCh38
rs1043900354 Chr.12: 76346890-76346890 on Build GRCh38
rs201335653 Chr.12: 76346957-76346957 on Build GRCh38
rs11109474 Chr.12: 76346859-76346859 on Build GRCh38
rs138702315 Chr.12: 76346827-76346827 on Build GRCh38
rs776775792 Chr.12: 76346856-76346856 on Build GRCh38
rs764234716 Chr.12: 76346760-76346760 on Build GRCh38
rs766313616 Chr.12: 76346868-76346868 on Build GRCh38
rs754911313 Chr.12: 76346882-76346882 on Build GRCh38
rs763827381 Chr.12: 76346817-76346818 on Build GRCh38
rs964776134 Chr.12: 76346774-76346774 on Build GRCh38
rs754502448 Chr.12: 76346772-76346772 on Build GRCh38
rs767572725 Chr.12: 76346910-76346910 on Build GRCh38
rs779201114 Chr.12: 76346931-76346931 on Build GRCh38
rs748933585 Chr.12: 76346913-76346913 on Build GRCh38
rs772592573 Chr.12: 76346837-76346837 on Build GRCh38
rs370079788 Chr.12: 76346793-76346793 on Build GRCh38

View Full Product Details