Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129619929-129620176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf50
Entrez Gene ID
375759
Gene Name
chromosome 9 open reading frame 50
Gene Aliases
-
UniGene
Hs.124223
Gene Chromosome Location
Chr.9: 129612225-129622275 on Build GRCh38
Transcripts
NM_199350.3 XM_011518655.1 XM_011518656.1 XM_011518657.2 XM_011518658.2 XM_011518659.1 XM_011518660.1 XM_011518661.2 XM_011518662.2 XM_011518663.1 XM_017014708.1 NM_001286796.1

Gene Symbol
NTMT1
Entrez Gene ID
28989
Gene Name
N-terminal Xaa-Pro-Lys N-methyltransferase 1
Gene Aliases
AD-003, C9orf32, HOMT1A, METTL11A, NRMT, NTM1A
UniGene
Hs.744027
Gene Chromosome Location
Chr.9: 129608884-129636742 on Build GRCh38
Transcripts
NM_199350.3 XM_011518655.1 XM_011518656.1 XM_011518657.2 XM_011518658.2 XM_011518659.1 XM_011518660.1 XM_011518661.2 XM_011518662.2 XM_011518663.1 XM_017014708.1 NM_001286796.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6391195 c.431G>T p.G144V Chr.9: 129620144-129620144 on Build GRCh38
6499047 c.455_475del21 p.R152_L158delREFLHQL Chr.9: 129620100-129620120 on Build GRCh38
5416898 c.507_508+17del19 p.? Chr.9: 129620050-129620068 on Build GRCh38
6407758 c.464T>C p.L155P Chr.9: 129620111-129620111 on Build GRCh38
6408674 c.401A>G p.E134G Chr.9: 129620174-129620174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367589276 Chr.9: 129620015-129620015 on Build GRCh38
rs945013269 Chr.9: 129620075-129620075 on Build GRCh38
rs962396918 Chr.9: 129619953-129619953 on Build GRCh38
rs548738466 Chr.9: 129620155-129620155 on Build GRCh38
rs1042805379 Chr.9: 129620134-129620134 on Build GRCh38
rs979850751 Chr.9: 129620042-129620042 on Build GRCh38
rs904304018 Chr.9: 129620142-129620142 on Build GRCh38
rs573286907 Chr.9: 129620053-129620053 on Build GRCh38
rs529361671 Chr.9: 129620130-129620130 on Build GRCh38
rs1024106548 Chr.9: 129620009-129620009 on Build GRCh38
rs111423646 Chr.9: 129620061-129620061 on Build GRCh38
rs1049161142 Chr.9: 129620133-129620133 on Build GRCh38
rs945754052 Chr.9: 129620064-129620064 on Build GRCh38
rs927184565 Chr.9: 129620016-129620016 on Build GRCh38
rs544557598 Chr.9: 129619970-129619970 on Build GRCh38
rs7467760 Chr.9: 129619974-129619974 on Build GRCh38
rs980792791 Chr.9: 129619987-129619987 on Build GRCh38
rs970296517 Chr.9: 129620021-129620021 on Build GRCh38
rs938595092 Chr.9: 129620022-129620022 on Build GRCh38
rs577950105 Chr.9: 129619979-129619979 on Build GRCh38
rs749921277 Chr.9: 129620098-129620098 on Build GRCh38
rs1041034371 Chr.9: 129620077-129620077 on Build GRCh38
rs560342192 Chr.9: 129620055-129620055 on Build GRCh38
rs762764772 Chr.9: 129620095-129620097 on Build GRCh38
rs1015501228 Chr.9: 129619978-129619978 on Build GRCh38
rs932159897 Chr.9: 129620118-129620118 on Build GRCh38
rs778348522 Chr.9: 129620082-129620082 on Build GRCh38
rs922085380 Chr.9: 129620080-129620080 on Build GRCh38
rs540670707 Chr.9: 129619983-129619983 on Build GRCh38
rs377328547 Chr.9: 129619980-129619980 on Build GRCh38

View Full Product Details