Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 84609450-84609723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDS1
Entrez Gene ID
1040
Gene Name
CDP-diacylglycerol synthase 1
Gene Aliases
CDS
UniGene
Hs.654899
Gene Chromosome Location
Chr.4: 84582904-84651340 on Build GRCh38
Transcripts
NM_001263.3 XM_005262687.2 XM_017007648.1 XM_017007649.1 XM_017007650.1 XM_017007651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
734431 c.306G>A p.L102L Chr.4: 84609489-84609489 on Build GRCh38
3393027 c.293C>T p.S98L Chr.4: 84609476-84609476 on Build GRCh38
7228256 c.297G>T p.L99F Chr.4: 84609480-84609480 on Build GRCh38
3606663 c.303C>T p.F101F Chr.4: 84609486-84609486 on Build GRCh38
3606664 c.309C>G p.I103M Chr.4: 84609492-84609492 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs17009168 Chr.4: 84609643-84609643 on Build GRCh38
rs915970157 Chr.4: 84609648-84609648 on Build GRCh38
rs79423189 Chr.4: 84609673-84609673 on Build GRCh38
rs571010495 Chr.4: 84609546-84609546 on Build GRCh38
rs920313104 Chr.4: 84609590-84609590 on Build GRCh38
rs762484703 Chr.4: 84609548-84609550 on Build GRCh38
rs373758079 Chr.4: 84609497-84609497 on Build GRCh38
rs779650106 Chr.4: 84609542-84609542 on Build GRCh38
rs562774914 Chr.4: 84609522-84609522 on Build GRCh38
rs1050640319 Chr.4: 84609605-84609605 on Build GRCh38
rs75287618 Chr.4: 84609571-84609571 on Build GRCh38
rs970566649 Chr.4: 84609677-84609677 on Build GRCh38
rs892397589 Chr.4: 84609668-84609668 on Build GRCh38
rs183570204 Chr.4: 84609492-84609492 on Build GRCh38
rs36068434 Chr.4: 84609480-84609480 on Build GRCh38
rs763623904 Chr.4: 84609491-84609491 on Build GRCh38
rs200859363 Chr.4: 84609507-84609507 on Build GRCh38
rs930387451 Chr.4: 84609599-84609599 on Build GRCh38
rs765468711 Chr.4: 84609487-84609487 on Build GRCh38
rs1032262203 Chr.4: 84609614-84609614 on Build GRCh38
rs750565728 Chr.4: 84609536-84609536 on Build GRCh38
rs1009328154 Chr.4: 84609690-84609690 on Build GRCh38
rs765230613 Chr.4: 84609613-84609613 on Build GRCh38
rs778795741 Chr.4: 84609553-84609553 on Build GRCh38
rs999392531 Chr.4: 84609585-84609585 on Build GRCh38
rs997161950 Chr.4: 84609517-84609517 on Build GRCh38
rs80223479 Chr.4: 84609595-84609595 on Build GRCh38
rs991069862 Chr.4: 84609645-84609645 on Build GRCh38
rs1040274353 Chr.4: 84609666-84609666 on Build GRCh38
rs755952224 Chr.4: 84609539-84609539 on Build GRCh38
rs754964163 Chr.4: 84609550-84609550 on Build GRCh38
rs375223136 Chr.4: 84609564-84609564 on Build GRCh38
rs201583321 Chr.4: 84609530-84609530 on Build GRCh38
rs888779989 Chr.4: 84609641-84609641 on Build GRCh38
rs751739087 Chr.4: 84609493-84609493 on Build GRCh38

View Full Product Details