Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 155073241-155073514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRCC3
Entrez Gene ID
79184
Gene Name
BRCA1/BRCA2-containing complex subunit 3
Gene Aliases
BRCC36, C6.1A, CXorf53
UniGene
Hs.558537
Gene Chromosome Location
Chr.X: 155071403-155126766 on Build GRCh38
Transcripts
NM_001018055.2 NM_001242640.1 NM_024332.3 XM_005274751.4 XM_017029838.1

SNP Detail

SNP ID Chromosome Location
rs781808889 Chr.X: 155073415-155073415 on Build GRCh38
rs371411951 Chr.X: 155073446-155073446 on Build GRCh38
rs782698211 Chr.X: 155073414-155073414 on Build GRCh38
rs376767171 Chr.X: 155073358-155073358 on Build GRCh38
rs782799702 Chr.X: 155073323-155073323 on Build GRCh38
rs782476658 Chr.X: 155073358-155073358 on Build GRCh38
rs985206068 Chr.X: 155073320-155073320 on Build GRCh38
rs781905422 Chr.X: 155073328-155073328 on Build GRCh38
rs781918754 Chr.X: 155073347-155073347 on Build GRCh38
rs782493159 Chr.X: 155073348-155073348 on Build GRCh38
rs782348088 Chr.X: 155073345-155073345 on Build GRCh38
rs782568261 Chr.X: 155073308-155073308 on Build GRCh38
rs782264929 Chr.X: 155073368-155073368 on Build GRCh38
rs782120431 Chr.X: 155073366-155073366 on Build GRCh38
rs782188634 Chr.X: 155073354-155073354 on Build GRCh38
rs782104034 Chr.X: 155073382-155073382 on Build GRCh38
rs782202712 Chr.X: 155073338-155073338 on Build GRCh38
rs1039542146 Chr.X: 155073412-155073412 on Build GRCh38
rs782265745 Chr.X: 155073338-155073338 on Build GRCh38
rs782434860 Chr.X: 155073331-155073331 on Build GRCh38
rs950708709 Chr.X: 155073309-155073309 on Build GRCh38
rs782757585 Chr.X: 155073470-155073470 on Build GRCh38
rs933886741 Chr.X: 155073437-155073437 on Build GRCh38
rs782037747 Chr.X: 155073359-155073359 on Build GRCh38
rs782636941 Chr.X: 155073329-155073329 on Build GRCh38
rs782306286 Chr.X: 155073375-155073375 on Build GRCh38
rs782723814 Chr.X: 155073347-155073348 on Build GRCh38

View Full Product Details