Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 25123898-25124165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYCS
Entrez Gene ID
54205
Gene Name
cytochrome c, somatic
Gene Aliases
CYC, HCS, THC4
UniGene
Hs.437060
Gene Chromosome Location
Chr.7: 25118651-25125361 on Build GRCh38
Transcripts
NM_018947.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6261352 c.54C>A p.C18* Chr.7: 25124066-25124066 on Build GRCh38
1088724 c.95A>G p.N32S Chr.7: 25124025-25124025 on Build GRCh38
6177284 c.4G>C p.G2R Chr.7: 25124116-25124116 on Build GRCh38
6339115 c.93A>T p.P31P Chr.7: 25124027-25124027 on Build GRCh38
1088725 c.29T>G p.I10S Chr.7: 25124091-25124091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563697201 Chr.7: 25124115-25124115 on Build GRCh38
rs11548815 Chr.7: 25124059-25124059 on Build GRCh38
rs543905568 Chr.7: 25124083-25124083 on Build GRCh38
rs121918552 Chr.7: 25123996-25123996 on Build GRCh38
rs565777563 Chr.7: 25123941-25123941 on Build GRCh38
rs775735347 Chr.7: 25124111-25124111 on Build GRCh38
rs11548802 Chr.7: 25124044-25124044 on Build GRCh38
rs11548805 Chr.7: 25124016-25124016 on Build GRCh38
rs11548772 Chr.7: 25123969-25123969 on Build GRCh38
rs769988994 Chr.7: 25124096-25124096 on Build GRCh38
rs752984039 Chr.7: 25124020-25124020 on Build GRCh38
rs573366932 Chr.7: 25123966-25123966 on Build GRCh38
rs774034189 Chr.7: 25124126-25124126 on Build GRCh38
rs11267038 Chr.7: 25124125-25124125 on Build GRCh38
rs11548796 Chr.7: 25124029-25124029 on Build GRCh38
rs758819864 Chr.7: 25123942-25123942 on Build GRCh38
rs930911491 Chr.7: 25123986-25123986 on Build GRCh38
rs146569499 Chr.7: 25124086-25124086 on Build GRCh38
rs17851278 Chr.7: 25124067-25124067 on Build GRCh38
rs1047899818 Chr.7: 25124004-25124004 on Build GRCh38
rs11548812 Chr.7: 25124000-25124000 on Build GRCh38
rs11548807 Chr.7: 25123990-25123990 on Build GRCh38
rs11548797 Chr.7: 25124048-25124048 on Build GRCh38
rs11548783 Chr.7: 25123978-25123978 on Build GRCh38
rs761747583 Chr.7: 25124131-25124131 on Build GRCh38
rs11548816 Chr.7: 25124061-25124061 on Build GRCh38
rs11548795 Chr.7: 25123953-25123953 on Build GRCh38
rs11548779 Chr.7: 25124027-25124027 on Build GRCh38
rs986520087 Chr.7: 25123955-25123955 on Build GRCh38
rs886037737 Chr.7: 25123975-25123975 on Build GRCh38
rs778031575 Chr.7: 25124060-25124060 on Build GRCh38
rs11548791 Chr.7: 25124005-25124005 on Build GRCh38
rs774382732 Chr.7: 25123977-25123977 on Build GRCh38
rs757774333 Chr.7: 25124081-25124081 on Build GRCh38
rs375916000 Chr.7: 25123973-25123973 on Build GRCh38
rs747501574 Chr.7: 25124063-25124063 on Build GRCh38
rs778394280 Chr.7: 25123948-25123948 on Build GRCh38
rs11548799 Chr.7: 25124035-25124035 on Build GRCh38
rs11548798 Chr.7: 25124043-25124043 on Build GRCh38
rs11548806 Chr.7: 25124057-25124057 on Build GRCh38
rs113106786 Chr.7: 25124082-25124082 on Build GRCh38
rs771002682 Chr.7: 25123963-25123963 on Build GRCh38
rs779357270 Chr.7: 25123939-25123939 on Build GRCh38
rs747030332 Chr.7: 25123962-25123962 on Build GRCh38
rs746031832 Chr.7: 25123967-25123967 on Build GRCh38
rs542089649 Chr.7: 25123979-25123979 on Build GRCh38
rs749961896 Chr.7: 25123997-25123997 on Build GRCh38
rs34522469 Chr.7: 25124121-25124122 on Build GRCh38
rs775258821 Chr.7: 25123968-25123968 on Build GRCh38

View Full Product Details