Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99713362-99713635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP3A7
Entrez Gene ID
1551
Gene Name
cytochrome P450 family 3 subfamily A member 7
Gene Aliases
CP37, CYPIIIA7, P-450(HFL33), P-450111A7, P450-HFLA
UniGene
Hs.111944
Gene Chromosome Location
Chr.7: 99705037-99735200 on Build GRCh38
Transcripts
NM_000765.4 NM_001256497.2

Gene Symbol
CYP3A7-CYP3A51P
Entrez Gene ID
100861540
Gene Name
CYP3A7-CYP3A51P readthrough
Gene Aliases
CYP3A7-3AP1, CYP3A7-CYP3AP1, CYP3A7.1L
UniGene
Gene Chromosome Location
Chr.7: 99684679-99735200 on Build GRCh38
Transcripts
NM_000765.4 NM_001256497.2

Gene Symbol
ZSCAN25
Entrez Gene ID
221785
Gene Name
zinc finger and SCAN domain containing 25
Gene Aliases
ZNF498
UniGene
Hs.446297
Gene Chromosome Location
Chr.7: 99616600-99738689 on Build GRCh38
Transcripts
NM_000765.4 NM_001256497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6111236 c.825G>A p.M275I Chr.7: 99713509-99713509 on Build GRCh38
6639314 c.801C>T p.H267H Chr.7: 99713533-99713533 on Build GRCh38
5345438 c.799-4C>A p.? Chr.7: 99713539-99713539 on Build GRCh38
7245164 c.842C>G p.S281* Chr.7: 99713492-99713492 on Build GRCh38
4828855 c.855G>A p.E285E Chr.7: 99713479-99713479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776972099 Chr.7: 99713449-99713449 on Build GRCh38
rs955842121 Chr.7: 99713612-99713612 on Build GRCh38
rs760750617 Chr.7: 99713574-99713575 on Build GRCh38
rs1015451532 Chr.7: 99713473-99713473 on Build GRCh38
rs779390865 Chr.7: 99713472-99713472 on Build GRCh38
rs770149947 Chr.7: 99713563-99713563 on Build GRCh38
rs575168337 Chr.7: 99713511-99713511 on Build GRCh38
rs748022481 Chr.7: 99713418-99713418 on Build GRCh38
rs747455500 Chr.7: 99713445-99713445 on Build GRCh38
rs767621705 Chr.7: 99713498-99713498 on Build GRCh38
rs769134726 Chr.7: 99713535-99713535 on Build GRCh38
rs762217964 Chr.7: 99713456-99713456 on Build GRCh38
rs577476945 Chr.7: 99713476-99713476 on Build GRCh38
rs201467317 Chr.7: 99713575-99713575 on Build GRCh38
rs200663057 Chr.7: 99713507-99713507 on Build GRCh38
rs754962953 Chr.7: 99713488-99713488 on Build GRCh38
rs912221165 Chr.7: 99713592-99713592 on Build GRCh38
rs745902810 Chr.7: 99713438-99713438 on Build GRCh38
rs772149342 Chr.7: 99713439-99713439 on Build GRCh38
rs368191881 Chr.7: 99713576-99713576 on Build GRCh38
rs773465910 Chr.7: 99713569-99713569 on Build GRCh38
rs200137477 Chr.7: 99713515-99713515 on Build GRCh38
rs367858678 Chr.7: 99713437-99713437 on Build GRCh38
rs575597575 Chr.7: 99713541-99713541 on Build GRCh38
rs772884241 Chr.7: 99713475-99713475 on Build GRCh38
rs115410773 Chr.7: 99713484-99713484 on Build GRCh38
rs143911062 Chr.7: 99713477-99713477 on Build GRCh38
rs780039093 Chr.7: 99713531-99713531 on Build GRCh38
rs769151350 Chr.7: 99713448-99713448 on Build GRCh38
rs563529139 Chr.7: 99713532-99713532 on Build GRCh38
rs45585039 Chr.7: 99713577-99713577 on Build GRCh38
rs183629375 Chr.7: 99713440-99713440 on Build GRCh38
rs759167195 Chr.7: 99713586-99713586 on Build GRCh38
rs758560391 Chr.7: 99713529-99713529 on Build GRCh38
rs748638876 Chr.7: 99713550-99713550 on Build GRCh38
rs1005364493 Chr.7: 99713468-99713468 on Build GRCh38
rs375632080 Chr.7: 99713566-99713566 on Build GRCh38

View Full Product Details